Skip to Content
Merck
All Photos(5)

Key Documents

HPA024621

Sigma-Aldrich

Anti-ADAM2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-ADAM 2, Anti-CT15, Anti-Cancer/testis antigen 15, Anti-Disintegrin and metalloproteinase domain-containing protein 2, Anti-Fertilin subunit beta, Anti-PH-30, Anti-PH30, Anti-PH30-beta

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

CENCLFMSKERMCRPSFEECDLPEYCNGSSASCPENHYVQTGHPCGLNQWICIDGVCMSGDK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM2(2515)

General description

The gene ADAM2 (disintegrin and metalloproteinase domain-containing protein 2) is mapped to human chromosome 8p11. It belongs to the ADAM family of proteins. The protein is present in the testis. It protein has a disintegrin-like domain.

Immunogen

Disintegrin and metalloproteinase domain-containing protein 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ADAM2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ADAM2 (disintegrin and metalloproteinase domain-containing protein 2) is an integrin binding protein. Absence of ADAM2 in mice causes defects in sperm-egg binding. In addition, it helps in transfer of sperm from the uterus into the oviduct.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76215

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Al Zeyadi et al.
Biotechnology, biotechnological equipment, 29(1), 111-118 (2015-05-29)
Lung cancer is a serious health problem, since it is one of the leading causes for death worldwide. Molecular-cytogenetic studies could provide reliable data about genetic alterations which could be related to disease pathogenesis and be used for better prognosis
Ulyana V Desiderio et al.
PloS one, 5(10), e13744-e13744 (2010-11-10)
Integrins are heterodimeric cell adhesion molecules, with 18 α (ITGA) and eight β (ITGB) subunits forming 24 heterodimers classified into five families. Certain integrins, especially the α(4)/α(9) (ITGA4/ITGA9) family, interact with members of the ADAM (a disintegrin and metalloprotease) family.
Heejin Choi et al.
PloS one, 11(6), e0158321-e0158321 (2016-06-25)
The members of the ADAM (a disintegrin and metalloprotease) family are membrane-anchored multi-domain proteins that play prominent roles in male reproduction. ADAM2, which was one of the first identified ADAMs, is the best studied ADAM in reproduction. In the male
Charles Gyamera-Acheampong et al.
Fertility and sterility, 93(4), 1112-1123 (2009-04-04)
To study the molecular basis for the accelerated capacitation rate in PCSK4-null sperm. Comparative and controlled experimental research study. Academic medical institute. Male mice C57BL/6J wild-type or null congenics for the Pcsk4 allele. Cauda and epididymal sperm were capacitated for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service