Skip to Content
Merck
All Photos(2)

Key Documents

HPA023886

Sigma-Aldrich

Anti-MYO1E antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myosin-Ic, Anti-Myosin-Ie

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VIRNQYVPYPHAPGSQRSNQKSLYTSMARPPLPRQQSTSSDRVSQTPESLDFLKVPDQGAAGVRRQTTSRPPPAGGRPKPQPKPKPQVPQCKALYAYDAQDTDELSFNANDIIDIIKEDPSGWWTGRLRGKQG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO1E(4643)

General description

Myosin 1E (Myo1E), also called as myr-3 or human myosin-IC, belongs to the subclass-1 of the myosin-I family. It is encoded by a gene mapped to human chromosome 15q21. Myosin 1E is a long-tailed protein, ubiquitously expressed in vertebrates . Wild-type Myo1E protein is localized to plasma membrane, but mutant Myo1E is expressed in the podocyte cytoplasm. Single headed, low molecular weight Myo1E protein consists of motor-head domain involved in binding ATP and F-actin, a calmodulin-binding neck domain and a tail domain that interacts with membranes and tail-binding proteins.

Immunogen

Myosin-Ie recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Myosin 1E (Myo1E) plays a vital role in clathrin-mediated endocytosis in HeLa cells and podocyte function in the kidney. It is also associated with proteins that take part in actin dynamics. Myo1E supports the function of glomerular filtration barrier in humans. Aberrations in the expression of this gene causes glomerulosclerosis. Members of myosin-I family help in membrane dynamics and membrane trafficking.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85179

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MYO1E mutations and childhood familial focal segmental glomerulosclerosis.
Mele C
The New England Journal of Medicine, 365(4), 295-306 (2011)
Myo1e binds anionic phospholipids with high affinity.
Feeser EA
Biochemistry, 49(43), 9353-9360 (2010)
The kinetic mechanism of Myo1e (human myosin-IC).
El Mezgueldi M
The Journal of Biological Chemistry, 277(24), 21514-21521 (2002)
Alaina H Willet et al.
Molecular biology of the cell, 34(11), br17-br17 (2023-08-02)
Myosin-1s are monomeric actin-based motors that function at membranes. Myo1 is the single myosin-1 isoform in Schizosaccharomyces pombe that works redundantly with Wsp1-Vrp1 to activate the Arp2/3 complex for endocytosis. Here, we identified Ank1 as an uncharacterized cytoplasmic Myo1 binding
Michael J Randles et al.
Matrix biology : journal of the International Society for Matrix Biology, 90, 61-78 (2020-03-10)
Cells have evolved mechanisms to sense the composition of their adhesive microenvironment. Although much is known about general mechanisms employed by adhesion receptors to relay signals between the extracellular environment and the cytoskeleton, the nuances of ligand-specific signalling remain undefined.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service