Skip to Content
Merck
All Photos(4)

Key Documents

HPA023597

Sigma-Aldrich

Anti-LRBA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Beige-like protein, Anti-CDC4-like protein, Anti-Lipopolysaccharide-responsive and beige-like anchor protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

GGWRVWVDTLSITHSKVTFEIHKENLANIFREQQGKVDEEIGLCSSTSVQAASGIRRDINVSVGSQQPDTKDSPVCPHFTTNGNENSSIEKTSSLESASNIELQTTNTSYEEMKAEQENQELPDEGTLEETL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRBA(987)

General description

The gene LRBA (lipopolysaccharide-responsive and beige-like anchor protein) is mapped to human chromosome 4q31.3. It belongs to the BEACH (beige and chediak-higashi)-WD40 (β-transducin) protein family. LRBA is ubiquitously expressed. The protein localizes in the cytoplasm. Upon activation of B cells, LRBA moves to the membrane of trans-Golgi network, lysosomes, endoplasmic reticulum and to the cell surface.

Immunogen

Lipopolysaccharide-responsive and beige-like anchor protein recombinant protein epitope signature tag (PrEST)

Application

Anti-LRBA antibody produced in rabbit has been used in western blotting.
All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The protein encoded by the gene LRBA (lipopolysaccharide-responsive and beige-like anchor protein) participates in endocytosis of ligand-activated receptors. Mutations in LRBA are linked with inflammatory bowel disease (IBD)-like symptoms, autoimmune lymphoproliferative syndrome-like disease and IPEX (immune dysregulation, polyendocrinopathy, enteropathy, X-linked) syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74668

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atypical Manifestation of LPS-Responsive Beige-Like Anchor Deficiency Syndrome as an Autoimmune Endocrine Disorder without Enteropathy and Immunodeficiency.
Bakhtiar S
Frontiers in pediatrics, 4 (2016)
Shahrzad Bakhtiar et al.
Frontiers in immunology, 8, 52-52 (2017-02-16)
Inflammatory bowel disease (IBD) in young children can be a clinical manifestation of various primary immunodeficiency syndromes. Poor clinical outcome is associated with poor quality of life and high morbidity from the complications of prolonged immunosuppressive treatment and malabsorption. In
Wen-Juan Tang et al.
World journal of clinical cases, 9(21), 5873-5888 (2021-08-10)
Patients with lipopolysaccharide (LPS)-responsive beige-like anchor protein (LRBA) deficiency have a variety of clinical symptoms, but there is no apparent genotype-phenotype correlation, and patients carrying the same mutations may have different phenotypes. Therefore, it is not easy for doctors to
Eriko Totsune et al.
Frontiers in immunology, 12, 677572-677572 (2021-04-30)
Lipopolysaccharide-responsive beige-like anchor (LRBA) deficiency is a subtype of common variable immune deficiency (CVID). Numerous case reports and cohort studies have described a broad spectrum of clinical manifestations and variable disease phenotypes, including immune dysregulation, enteropathy, and recurrent infections. Although
Fabienne Charbit-Henrion et al.
Journal of Crohn's & colitis (2018-05-23)
An expanding number of monogenic defects have been identified as causative of severe forms of very early-onset inflammatory bowel diseases (VEO-IBD). The present study aimed at defining how next-generation sequencing (NGS) methods can be used to improve identification of known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service