Skip to Content
Merck
All Photos(7)

Key Documents

HPA021032

Sigma-Aldrich

Anti-HSD17B1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-17-beta-HSD 1, Anti-17-beta-hydroxysteroid dehydrogenase type 1, Anti-20 alpha-hydroxysteroid dehydrogenase, Anti-20-alpha-HSD, Anti-E2DH, Anti-Estradiol 17-beta-dehydrogenase 1, Anti-Placental 17-beta-hydroxysteroid dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FGVHLSLIECGPVHTAFMEKVLGSPEEVLDRTDIHTFHRFYQYLAHSKQVFREAAQNPEEVAEVFLTALRAPKPTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSD17B1(3292)

General description

The gene HSD17B14 (17-β-hydroxysteroid dehydrogenase 14) is mapped to human chromosome 19q13.33. It belongs to 17β hydroxysteroid dehydrogenases (17βHSDs) family of proteins. HSD17B14 is strongly expressed in the brain, liver and placenta. The protein localizes in the cytoplasm.

Immunogen

Estradiol 17-beta-dehydrogenase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

17β hydroxysteroid dehydrogenases (17βHSDs) are mainly involved in oxidation/reduction at position C17 of sex steroids. HSD17B14 oxidizes estradiol and testosterone into less bioactive steroid metabolites. Humans with high levels of HSD17B14 have lower local recurrences of estrogen positive breast cancer when treated with tamoxifen.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75157

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Sivik et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 44(13), 949-956 (2012-08-07)
17βHSD enzymes catalyze the stereospecific oxidation/reduction at carbon 17β of androgens and estrogens, and are important players in intracrine sex hormone synthesis. The biological relevance of 17βHSD14, first named retSDR3, is largely unknown. We generated and validated an antibody targeting
Germline BAX deletion in a patient with melanoma and gastrointestinal stromal tumor.
Amanda G Silva et al.
The American journal of gastroenterology, 108(8), 1372-1375 (2013-08-06)
Luis Z Blanco et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 30(4), 563-576 (2017-01-07)
Epithelial ovarian tumors are responsive to steroid hormone stimulation and the ovarian stroma may have a direct role in this process. We evaluated immunohistochemical markers of sex-steroid differentiation and steroidogenesis (calretinin, inhibin, steroidogenic factor 1), steroid enzymes involved in hormone
Tove Sivik et al.
PloS one, 7(7), e40568-e40568 (2012-07-14)
17β-Hydroxysteroid dehydrogenases (17βHSDs) are important enzymes regulating the pool of bioactive steroids in the breast. The current study was undertaken in order to evaluate implications of 17βHSD14 in breast cancer, measuring 17βHSD14 protein expression in breast tumours. An antibody targeting
Petra Lukacik et al.
The Biochemical journal, 402(3), 419-427 (2006-10-28)
To this day, a significant proportion of the human genome remains devoid of functional characterization. In this study, we present evidence that the previously functionally uncharacterized product of the human DHRS10 gene is endowed with 17beta-HSD (17beta-hydroxysteroid dehydrogenase) activity. 17beta-HSD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service