Skip to Content
Merck
All Photos(3)

Key Documents

HPA020735

Sigma-Aldrich

Anti-GGCT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cytochrome c-releasing factor 21, Anti-Gamma-glutamylcyclotransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

QEGVKSGMYVVIEVKVATQEGKEITCRSYLMTNYESAPPSPQYKKIICMGAKENGLPLEYQEKLKAIEPNDYTGKVSEEIE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGCT(79017)

General description

γ-glutamylcyclotransferase (GGCT) is a homodimeric protein. The gene encoding it is localized to human chromosome 7p14-15.

Immunogen

Gamma-glutamylcyclotransferase recombinant protein epitope signature tag (PrEST)

Application

Anti-GGCT antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

γ-glutamylcyclotransferase (GGCT) takes part in glutathione metabolism. It plays an important role in forming 5-oxoproline and free amino acids from γ-glutamyl peptides. This protein has been shown to be expressed in many cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74840

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhifeng Lin et al.
Cancer biotherapy & radiopharmaceuticals, 30(5), 211-216 (2015-05-06)
Lung cancer as an aggressive type tumor is rapidly growing and has become the leading cause of cancer-related death worldwide. γ-Glutamylcyclotransferase (GGCT) has been shown as a diagnostic marker in various cancers. To reveal whether there is a correlation between
Susumu Kageyama et al.
BioMed research international, 2015, 345219-345219 (2015-09-05)
Gamma-glutamylcyclotransferase (GGCT) is one of the major enzymes involved in glutathione metabolism. However, its gene locus was unknown for many years. Recently, the gene for GGCT was found to be identical to C7orf24, which is registered as a hypothetical protein.
Wenjie Zhang et al.
BMC biochemistry, 17(1), 19-19 (2016-12-03)
Gamma glutamylcyclotransferase (GGCT) has been proved to be involved in various cancers, but the biological function of GGCT in gastric cancer is still largely unknown. The expression level of GGCT was evaluated by informatics analyses based on the Oncomine database.
Tomonari Amano et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 60(1), 76-86 (2011-12-30)
γ-Glutamyl cyclotransferase (GGCT) contributes to the γ-glutamyl cycle that regulates glutathione metabolism. Although GGCT has been implicated in several studies as a possible cancer marker, little is known about its distribution in cells and tissues. The authors investigated GGCT expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service