Skip to Content
Merck
All Photos(6)

Key Documents

HPA020731

Sigma-Aldrich

Anti-GFRA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GDNF family receptor alpha-3, Anti-GFR-alpha-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LPTESRLMNSCLQARRKCQADPTCSAAYHHLDSCTSSISTPLPSEEPSVPADCLEAAQQLRNSSLIGCMCHRRMKNQVACL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GFRA3(2676)

General description

GDNF family receptor α-3 (GFRA3) is expressed in the nervous system. It belongs to the GDNF receptor-alpha (GFR-α) family of proteins. GFRA3 is made up of 400 amino acids and the gene encoding it is localized on human chromosome 5q31.

Immunogen

GDNF family receptor alpha-3 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GDNF family receptor α-3 (GFRA3) forms a ternary signaling complex with RET receptor and binds to the ligand, artemin (ARTN). The expression of this protein has been linked to pancreatic carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75162

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C I Onochie et al.
Journal of medical genetics, 37(9), 674-679 (2000-09-09)
The GDNF family receptor alpha (GFRalpha) proteins are extracellular cell surface bound molecules that act as adapters in binding of the GDNF family of soluble neurotrophic factors to the RET receptor. These molecules are essential for development of many neural
Dong-Liang Zhu et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 29(3), 428-432 (2009-03-24)
To investigate the association of artemin and GFRalpha3 expressions with perineural invasion and metastasis of pancreatic carcinoma. Semi-quantitative reverse transcription polymerase chain reaction (RT-PCR) and immunohistochemistry were used to detect the expression of artemin and GFRalpha3 in pancreatic carcinoma tissues
Xinquan Wang et al.
Structure (London, England : 1993), 14(6), 1083-1092 (2006-06-13)
Artemin (ARTN) is a member of the glial cell line-derived neurotrophic factor (GDNF) family ligands (GFLs) which regulate the development and maintenance of many neuronal populations in the mammalian nervous system. Here we report the 1.92 A crystal structure of
S Masure et al.
European journal of biochemistry, 251(3), 622-630 (1998-03-07)
Neurturin and glial-cell-line-derived neurotrophic factor (GDNF) promote the survival and maintenance of different types of neuronal cells and signal through a receptor complex composed of a ligand binding subunit, either GDNF family receptor alpha-1 (GFRalpha-1) or alpha-2 (GFRalpha-2), together with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service