Skip to Content
Merck
All Photos(9)

Key Documents

HPA019154

Sigma-Aldrich

Anti-SMAD4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deletion target in pancreatic carcinoma 4, Anti-Mothers against DPP homolog 4, Anti-Mothers against decapentaplegic homolog 4, Anti-SMAD 4, Anti-hSMAD4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AFDLKCDSVCVNPYHYERVVSPGIDLSGLTLQSNAPSSMMVKDEYVHDFEGQPSLSTEGHSIQTIQHPPSNRASTETYSTPALLAPSESNATSTANFPNIPVASTSQPASILGGSHSEGLLQI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD4(4089)

General description

The gene SMAD4 (mothers against decapentaplegic homolog 4) is mapped to human chromosome 18q21.1. The protein localizes in the cytoplasm and nucleus. SMAD4 is also referred as DPC4 (deleted in pancreatic carcinoma).

Immunogen

Mothers against decapentaplegic homolog 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SMAD4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Upon TGFβ (transforming growth factor β) activation, SMAD (mothers against decapentaplegic homolog) proteins are responsible for transcription activation in the nucleus. SMAD4 is a crucial protein of TGFβ signaling. It promotes association of SMAD2/4 to DNA and helps SMAD1/2 in transcription stimulation. Mutations in SMAD4 are linked with juvenile polyposis syndrome, hereditary haemorrhagic telangiectasia and Myhre syndrome. In renal cell carcinoma, it activates forkhead box protein H1 and thereby inhibits the progression of the carcinoma. SMAD4 is considered as a tumor suppressor protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86689

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunli Liu et al.
Molecular medicine reports, 11(4), 2717-2722 (2014-12-09)
Smad4 has recently been identified as a tumor suppressor gene in a variety of cancers, yet the role of Smad4 in renal cell carcinoma (RCC) remained to be elusive. Therefore, the aim of the present study was to explore the
Lina Jia et al.
Molecular cancer research : MCR, 12(12), 1779-1795 (2014-07-26)
SMAD4 has been suggested to inhibit the activity of the WNT/β-catenin signaling pathway in cancer. However, the mechanism by which SMAD4 antagonizes WNT/β-catenin signaling in cancer remains largely unknown. Aurora A kinase (AURKA), which is frequently overexpressed in cancer, increases
R Houlston et al.
Human molecular genetics, 7(12), 1907-1912 (1998-11-13)
Juvenile polyps are present in a number of Mendelian disorders, sometimes in association only with gastrointestinal cancer [juvenile polyposis syndrome (JPS)] and sometimes as part of known syndromes (Cowden, Gorlin and Banayan-Zonana) in association with developmental abnormalities, dysmorphic features or
Jochen K M Lennerz et al.
The American journal of pathology, 179(2), 1015-1029 (2011-06-28)
Cirrhotic septa harbor vessels and inflammatory, fibrogenic, and ductular epithelial cells, collectively referred to as the ductular reaction (DR). Lack of the DR in the stromal compartment around hepatocellular carcinoma (HCC) has been documented; however, the relationship of epithelial keratin
Hyun-A Seong et al.
The Journal of biological chemistry, 282(16), 12272-12289 (2007-03-01)
We have reported previously that PDK1 physically interacts with STRAP, a transforming growth factor-beta (TGF-beta) receptor-interacting protein, and enhances STRAP-induced inhibition of TGF-beta signaling. In this study we show that PDK1 coimmunoprecipitates with Smad proteins, including Smad2, Smad3, Smad4, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service