Skip to Content
Merck
All Photos(8)

Documents

HPA018849

Sigma-Aldrich

Anti-EVL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ena/VASP-like protein, Anti-Ena/vasodilator-stimulated phosphoprotein- like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SSQRQVQNGPSPDEMDIQRRQVMEQHQQQRQESLERRTSATGPILPPGHPSSAASAPVSCSGPPPPPPPPVPPPPTGATPPP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EVL(51466)

General description

The gene EVL (Ena/VASP-like protein) is mapped to human chromosome 14q32.2. It belongs to Ena/VASP family of proteins. The protein has three domains, EVH1 (Ena/VASP homology), proline-rich and EVH2. The rat EVL homolog is present in brain, lung, spleen, thymus and testis.

Immunogen

Ena/VASP-like protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ena/VASP protein family is involved in actin dynamics by associating with actin. Ena/VASP-like protein (EVL) interacts with lamellipodin and localizes to the tips of lamellipodia and filopodia to regulate cytoskeletal dynamics. EVL interacts with RAD51 (DNA repair protein) and RAD51B, and participates in homologous recombination repair of DNA double-strand breaks. It is down-regulated in colorectal adenocarcinomas and adenomas. On the other hand, it is up-regulated in breast tumors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74608

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumiaki Sato et al.
The FEBS journal, 278(10), 1598-1609 (2011-03-15)
MicroRNAs (miRNAs) comprise species of short noncoding RNA that regulate gene expression post-transcriptionally. Recent studies have demonstrated that epigenetic mechanisms, including DNA methylation and histone modification, not only regulate the expression of protein-encoding genes, but also miRNAs, such as let-7a
Motoki Takaku et al.
The Journal of biological chemistry, 284(21), 14326-14336 (2009-03-31)
The RAD51 protein is a central player in homologous recombinational repair. The RAD51B protein is one of five RAD51 paralogs that function in the homologous recombinational repair pathway in higher eukaryotes. In the present study, we found that the human
W M Grady et al.
Oncogene, 27(27), 3880-3888 (2008-02-12)
MicroRNAs are small, non-coding RNAs that influence gene regulatory networks by post-transcriptional regulation of specific messenger RNA targets. MicroRNA expression is dysregulated in human malignancies, frequently leading to loss of expression of certain microRNAs. We report that expression of hsa-miR-342
Motoki Takaku et al.
The FEBS journal, 276(20), 5841-5848 (2009-09-04)
EVL is a member of the ENA/VASP family, which is involved in actin-remodeling processes. Previously, we reported that human EVL directly interacts with RAD51, which is an essential protein in the homologous recombinational repair of DNA double-strand breaks, and stimulates
Motoki Takaku et al.
Nucleic acids research, 38(21), 7579-7586 (2010-07-20)
The human Ena/Vasp-like (EVL) protein is considered to be a bifunctional protein, involved in both actin remodeling and homologous recombination. In the present study, we found that human EVL forms heat-stable multimers of circular single-stranded DNA (ssDNA) molecules in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service