Skip to Content
Merck
All Photos(2)

Key Documents

HPA012530

Sigma-Aldrich

Anti-LGR5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

LGR5 Antibody - Anti-LGR5 antibody produced in rabbit, Lgr5 Antibody, Anti-G-protein coupled receptor 49, Anti-G-protein coupled receptor 67, Anti-Leucine-rich repeat-containing G-protein coupled receptor 5 precursor, Anti-Orphan G-protein coupled receptor HG38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

AIIHPNAFSTLPSLIKLDLSSNLLSSFPITGLHGLTHLKLTGNHALQSLISSENFPELKVIEMPYAYQCCAFGVCENAYKISNQWNKGDNSSMDDLHKKDAGMFQAQDERDLEDFLLDFEEDLKALHSVQCSPSPGPFKPCEHLLDG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LGR5(8549)

General description

Leucine-rich repeat containing G protein-coupled receptor 5 (LGR5) is a seven-transmembrane G-protein coupled receptor. It belongs to the family of glycoprotein hormone receptor family. LGR5 is present in the endometrium layer. The LGR5 gene is located on human chromosome location 12q21.1.

Immunogen

Leucine-rich repeat-containing G-protein coupled receptor 5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-LGR5 antibody produced in rabbit has been used in:
  • immunocytochemistry
  • immunohistochemistry
  • western blotting

Biochem/physiol Actions

Leucine-rich repeat containing G protein-coupled receptor 5 (LGR5) binds to R-spondin and enhances the Wnt/ β-catenin pathway. It stimulates tumor progression and induces colorectal cancer development. Lgr5 acts as a potential stem cell marker in numerous adult tissues. It has clinicopathological significance in various cancers, like colon, stomach, liver and esophagus. Elevated expression of LGR5 is associated with ovarian, colon and hepatocellular cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71276

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canine Epithelial Skin Tumours: Expression of the Stem Cell Markers Lgr5, Lgr6 and Sox9 in Light of New Cancer Stem Cell Theories
Bongiovanni L, et al.
Veterinary sciences, 7(2), 62-62 (2020)
Prognostic impact of mRNA levels of LGR5 transcript variants in OSCC patients
Rot S, et al.
BMC Cancer, 19(1), 155-155 (2019)
Filiz Mericli et al.
Pharmaceutical biology, 55(1), 1239-1248 (2017-03-07)
Almond oil is used in traditional and complementary therapies for its numerous health benefits due to high unsaturated fatty acids content. This study investigated the composition and in vitro anticancer activity of almond oil from Northern Cyprus and compared with
Eda Becer et al.
Gene, 686, 213-219 (2018-11-21)
Colchicum pusillum belongs to the family Colchicaceae that particularly rich in tropolonic alkaloids. The aim of this study was to investigate the cytotoxicity and in vitro anticancer activity of Colchicum pusillum ethanolic extract on Colo-320 primer and Colo-741 metastatic colon
Lgr5 Does Not Vary Throughout the Menstrual Cycle in Endometriotic Human Eutopic Endometrium
Vallve-Juanico J, et al.
International Journal of Molecular Sciences, 20(1), 22-22 (2019)

Articles

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service