Skip to Content
Merck
All Photos(4)

Key Documents

HPA012016

Sigma-Aldrich

Anti-ERBB4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALS19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
10
technique(s):
immunohistochemistry: 1:50- 1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LPSPNDSKFFQNLLDEEDLEDMMDAEEYLVPQAFNIPPPIYTSRARIDSNRNQFVYRDGGFAAEQGVSVPYRAPTSTIPEAPVAQGATAEIFDDSCCNGTLRK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERBB4(2066)

General description

ERBB4 (erb-b2 receptor tyrosine kinase 4) is a tissue-specific, tyrosine kinase receptor belonging to the epidermal growth factor receptor (EGFR) subfamily of receptor tyrosine kinases. It exists in two isoform, which are expressed mainly in the neural tissues such as cerebellum. Its expression also has been found in the kidney and heart. The functional activity of the protein is controlled by the neuregulins (NRG), betacellulin (BTC), and heparin-binding EGF-like growth factor.

Immunogen

Receptor tyrosine-protein kinase erbB-4 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ERBB4 (erb-b2 receptor tyrosine kinase 4) is involved in milk production and anabolic processes in normal mammary epithelia and cancer by controlling the YAP/Hippo pathway and mevalonate pathways. Its phosphatidyl inositol 3-kinase (PI3-K) binding site interacts with several EGF family members including neuregulins (NRG) for cellular responses related to cell proliferation, invasion, and cell growth suppression. Mutation in the kinase domain of ERBB4 is responsible for the malignant cancer cell growth. ERBB4 identifies oncogenic somatic mutations in high-risk early breast cancer, which proves its prognostic marker quality. Mutation in the gene also causes a neurological disorder, amyotrophic lateral sclerosis (ALS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70281

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guodong Han et al.
Journal of cellular biochemistry (2019-02-13)
microRNAs are involved in the control of cell growth and apoptosis; they also play an essential role in resistance towards trastuzumab, in breast cancer. The objective of this study was to identify differentially expressed microRNA(s) and explore its therapeutic role
Convergent and divergent cellular responses by ErbB4 isoforms in mammary epithelial cells.
Wali VB, Haskins JW, Gilmore-Hebert M, et al.
Molecular Cancer Research, 12(8), 1140-1155 (2014)
Kari J Kurppa et al.
PloS one, 9(7), e102388-e102388 (2014-07-19)
A number of genetic variants have been linked to increased risk of breast cancer. Little is, however, known about the prognostic significance of hereditary factors. Here, we investigated the frequency and prognostic significance of two ERBB4 promoter region variants, -782G>T
Yuji Takahashi et al.
American journal of human genetics, 93(5), 900-905 (2013-10-15)
Amyotrophic lateral sclerosis (ALS) is a devastating neurological disorder characterized by the degeneration of motor neurons and typically results in death within 3-5 years from onset. Familial ALS (FALS) comprises 5%-10% of ALS cases, and the identification of genes associated
Xiaoting Liang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(3), 4559-4570 (2018-12-20)
The age-related functional exhaustion limits potential efficacy of mesenchymal stem cells (MSC) in treating cardiovascular disease. Therefore, rejuvenation of aged MSC in the elderly population is of great interest. We have previously reported that Erb-B2 receptor tyrosine kinase 4 (

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service