Skip to Content
Merck
All Photos(6)

Key Documents

HPA010638

Sigma-Aldrich

Anti-GOLM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Golgi membrane protein 1, Anti-Golgi membrane protein GP73, Anti-Golgi phosphoprotein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

GKGNVLGNSKSQTPAPSSEVVLDSKRQVEKEETNEIQVVNEEPQRDRLPQEPGREQVVEDRPVGGRGFGGAGELGQTPQVQAALSVSQENPEMEGPERDQLVIPDGQEEEQEAAGEGRNQQKLRGEDDYNMDENEAESE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

GOLM1 (golgi membrane protein 1) gene is localized to human chromosome 9q21.33 and is expressed normal epithelial cells of tissues, such as the gut, prostate, kidneys, lungs and within the central nervous system. Its expression is observed to be upregulated in liver diseases and in certain cancers such as hepatocellular carcinoma (HCC), prostate cancer, and renal cell cancer. The protein contains a short N-terminal cytoplasmic domain, followed by a transmembrane domain (TMD). It also contains a larger C-terminal domain that faces the Golgi lumen. This domain facing the lumen has a coiled-coil domain and an acid tail capable of mediating protein–protein interactions. The protein has a molar mass of 73kDa and is found to be upregulated by viral infection.

Immunogen

Golgi membrane protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GOLM1 (golgi membrane protein 1) gene encodes a type-II Golgi transmembrane protein that is also called as Golgi phosphoprotein 2. It may function in epithelial cells and carcinogenesis. It may be involved in the transport of protein cargo through the Golgi apparatus. This protein is found to interact with secretory clusterin, a sulfated glycoprotein associated with cellular processes such as carcinogenesis, progression of cancer and stress response. It may serve as a predictive marker in hepatocellular carcinoma (HCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71956

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gehan H Ibrahim et al.
Molecular biology reports, 40(12), 7069-7075 (2013-11-05)
Hepatocellular carcinoma (HCC) incidence is fast-growing especially in countries highly prevalent with viral hepatitis. Its poor prognosis has driven the research toward the discovery of sensitive markers for early detection. We investigated the usefulness of serum Transforming growth factor-beta1 (TGF-β1)
R D Kladney et al.
Gene, 249(1-2), 53-65 (2000-06-01)
We report the isolation and characterization of GP73, a novel 73kDa human Golgi protein. The GP73 cDNA was cloned by differential screening of a cDNA library derived from the liver of a patient with adult giant-cell hepatitis (GCH), a rare
Vasileios Exarchos et al.
Frontiers in cardiovascular medicine, 9, 953582-953582 (2022-10-25)
Thrombogenicity remains a major issue in cardiovascular implants (CVIs). Complete surficial coverage of CVIs by a monolayer of endothelial cells (ECs) prior to implantation represents a promising strategy but is hampered by the overall logistical complexity and the high number
Christian Cedric Bongolo et al.
Stem cells international, 2022, 9748714-9748714 (2022-12-20)
Hepatocellular carcinoma (HCC) remains a significant health burden to date. Its early diagnosis and treatment are complicated by the lack of early diagnosis markers and multidrug resistance. microRNA regulation of HCC oncogenes are among the new diagnostic and therapeutic strategies
Yan Zhou et al.
Molecular biology reports, 38(3), 1457-1462 (2010-09-16)
Golgi phosphoprotein 2 (GOLPH2/GP73/GOLM1), a type-II Golgi transmembrane protein of unknown function, is up-regulated in many cancers. Its Golgi luminal domain is potentially the major functional domain. The goal of this study is to identify the proteins interacting with GOLPH2.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service