Skip to Content
Merck
All Photos(6)

Key Documents

HPA010008

Sigma-Aldrich

Anti-MTM1 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myotubularin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KFNVDGWTVYNPVEEYRRQGLPNHHWRITFINKCYELCDTYPALLVVPYRASDDDLRRVATFRSRNRIPVLSWIHPENKTVIVRCSQPLVGMSGKRNKDDEKYLDVIRETNKQISKLTIYDARPSVNAVANK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTM1(4534)

Looking for similar products? Visit Product Comparison Guide

General description

MTM1 (myotubularin 1) is a lipid phosphatase, which is abundant at the junctional SR (sarcoplasmic reticulum) of the muscle triads. It is a member of conserved phosphatase family myotubularin, which contains both active phosphatases and inactive myotubularin-related proteins (MTMRs).

Immunogen

Myotubularin recombinant protein epitope signature tag (PrEST)

Application

Anti-MTM1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

MTM1 (myotubularin 1) protein plays an essential role in facilitating the curvature of SR (sarcoplasmic reticulum) membrane and SR remodelling. Therefore, it is critical for homeostasis of calcium and excitation-contraction coupling. The major function of this protein is the dephosphorylation of phosphatidylinositol-3-phosphate (PtdIns3P) and phosphatidylinositol-3,5-bisphosphate (PtdIns(3,5P)2). Mutations in this gene are linked to X-linked myotubular myopathy (XLMTM), which is an inherited disorder characterized by severe generalized hypotonia and skeletal muscles weakness with respiratory insufficiency. Mostly this disease is fatal in the initial months of life. Heterozygous mutations resulting in abnormal splicing in this gene are associated with early onset, slowly progressive myopathy. c.431delT (p.Leu144fs) mutation in this gene results in bilateral subdural hygromas and cephalhaematomas in male twins suffering from severe myotubular myopathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74421

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vandana A Gupta et al.
PLoS genetics, 9(6), e1003583-e1003583 (2013-07-03)
X-linked myotubular myopathy (XLMTM) is a congenital disorder caused by mutations of the myotubularin gene, MTM1. Myotubularin belongs to a large family of conserved lipid phosphatases that include both catalytically active and inactive myotubularin-related proteins (i.e., "MTMRs"). Biochemically, catalytically inactive
Bilateral subdural hygromas and cephalhaematomas in male twins with severe myotubular myopathy caused by a Novel c.431delT (p.Leu144fs) mutation in MTM1 gene.
Tejasvi Chaudhari et al.
Journal of paediatrics and child health, 47(1-2), 64-65 (2010-05-27)
Carola Hedberg et al.
Neuromuscular disorders : NMD, 22(3), 244-251 (2011-11-22)
We have investigated a woman and her daughter with an early onset, slowly progressive myopathy. Muscle biopsy showed in both cases severe atrophy with marked fatty replacement. Frequent fibers with internalized nuclei were present but no typical features of centronuclear
Carlo Cosimo Campa et al.
Nature chemical biology, 14(8), 801-810 (2018-06-20)
Directional transport of recycling cargo from early endosomes (EE) to the endocytic recycling compartment (ERC) relies on phosphatidylinositol 3-phosphate (PtdIns(3)P) hydrolysis and activation of the small GTPase Rab11. However, how these events are coordinated is yet unclear. By using a
Pankaj B Agrawal et al.
American journal of human genetics, 95(2), 218-226 (2014-08-05)
Centronuclear myopathies (CNMs) are characterized by muscle weakness and increased numbers of central nuclei within myofibers. X-linked myotubular myopathy, the most common severe form of CNM, is caused by mutations in MTM1, encoding myotubularin (MTM1), a lipid phosphatase. To increase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service