Skip to Content
Merck
All Photos(8)

Key Documents

HPA008736

Sigma-Aldrich

Anti-DAXX antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Daxx antibody produced in rabbit, Anti-Death domain-associated protein 6 antibody produced in rabbit, Anti-EAP1 antibody produced in rabbit, Anti-ETS1-associated protein 1 antibody produced in rabbit, Anti-Fas death domain-associated protein antibody produced in rabbit, Anti-hDaxx antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
RNAi knockdown
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DEEEEAAAGKDGDKSPMSSLQISNEKNLEPGKQISRSSGEQQNKGRIVSPSLLSEEPLAPSSIDAESNGEQPEELTLEEESPVSQLFELEIEALPLDTPSSVETDISSSRKQSEEPFTTVLENGAGMVSSTSFNGGVSPHNWGDSG

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DAXX(1616)

General description

DAXX (death-domain associated protein) is a nuclear protein, which is highly conserved in nature. It shuttles between nucleus and cytoplasm. It was originally identified as a FAS-interacting protein, which induces FAS-mediated apoptosis. This gene is localized to human chromosome 6p21.3 in the MHC (major histocompatibility complex) locus. The encoded protein contains FAS-binding region in its C-terminal. It contains two putative nuclear localization signals.

Immunogen

Death domain-associated protein 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-DAXX antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

DAXX (death-domain associated protein) plays essential roles in control of transcription and viral infection and carcinogenesis. It controls multiple signaling pathways involved in cell survival and death. It forms a constitutive constituent of promyleocytic leukaemia protein (PML) nuclear bodies (PML-NBs). In human papillomavirus (HPV)-induced cervical carcinoma, this protein translocates gradually from nucleus to nuclear membrane to cytoplasm to plasma membrane. It acts as a bifunctional protein at centrosomic/pericentrosomic heterochromatin, where it controls the loading of heat shock protein H3.3 under normal conditions and the protection epigenetic identity of heterochromatin during stress conditions. The subcellular localization of this protein determines the sensitivity of macrophages to oxidized low density lipoprotein (ox-LDL), where its nuclear localization can negate the effect of ox-LDL.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86890

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Daxx enigma.
Michaelson J S
Apoptosis, 5(3), 217-220 (2000)
Shuang-yang Tang et al.
Diagnostic pathology, 9, 1-1 (2014-01-09)
To provide the basis for further exploring the effect and its mechanism of Death domain associated protein (Daxx) on the progress of cervical carcinoma induced by human papillomavirus (HPV), the distribution and location of Daxx in cervical carcinoma with high
Guozuo Xiong et al.
Molecular biology reports, 41(11), 7183-7190 (2014-08-15)
Here we aimed to evaluate the effects of DAXX subcellular localization on ox-LDL induced macrophages apoptosis. Cytoplasmic localization vector DAXX-W621A and nuclear localization vector DAXX-S667A were constructed by point mutation in DAXX. Blank vector, full length DAXX, DAXX-W621A, DAXX-S667A was
Christian Geis et al.
The Journal of surgical research, 194(2), 388-393 (2014-12-03)
Neuroendocrine tumors (NETs) of the ileum are sporadic tumors derived from submucosal gastrointestinal stem cells. They often show clinical symptoms only after hepatic metastasation when curative therapy is limited or impossible. In this study, we analyzed the expression of the
M Kiriakidou et al.
DNA and cell biology, 16(11), 1289-1298 (1997-12-24)
Murine Daxx, a protein that binds to Fas and enhances Fas-mediated apoptosis through a signal transduction pathway involving the Jun amino-terminal kinase, was recently described. Here we report the cloning of human and monkey Daxx cDNAs, the widespread expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service