Skip to Content
Merck
All Photos(5)

Key Documents

HPA008208

Sigma-Aldrich

Anti-NAPG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gamma-soluble NSF attachment protein, Anti-N-ethylmaleimide-sensitive factor attachment protein gamma, Anti-SNAP-gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RGRRFDEAALSIQKEKNIYKEIENYPTCYKKTIAQVLVHLHRNDYVAAERCVRESYSIPGFNGSEDCAALEQLLEGYDQQDQDQVSDVCNSPLFKYMDNDYAKLGLSLVVPGGGIKKKSPATPQAKPDGVTATAA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAPG(8774)

General description

The gene encoding N-ethylmaleimide-sensitive factor attachment protein gamma (NAPG) is present on chromosome 18p11.

Immunogen

Gamma-soluble NSF attachment protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

N-ethylmaleimide-sensitive factor attachment protein γ (NAPG) has an important role in cellular processes which is required for neurotransmission in the central nervous system. It aids the N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes hence helping in the process of Golgi transport.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71574

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S W Whiteheart et al.
Nature, 362(6418), 353-355 (1993-03-25)
The soluble NSF attachment proteins (SNAPs) enable N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes. Here we report the cloning and sequencing of complementary DNAs encoding alpha-, beta- and gamma-SNAPs. Two of these proteins, alpha and gamma, are found
Andrew E Weller et al.
Psychiatric genetics, 16(1), 3-8 (2006-01-06)
A number of studies have implicated the chromosome 18p11 region as a susceptibility region for bipolar disorder. The gene encoding gamma-SNAP (NAPG), one of three soluble N-ethylmaleimide-sensitive fusion (NSF)-attachment proteins (SNAPs), is located in the 18p11 region and is thought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service