Skip to Content
Merck
All Photos(11)

Key Documents

HPA008066

Sigma-Aldrich

Anti-CALD1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDM antibody produced in rabbit, Anti-Caldesmon antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SLKIEERAEFLNKSVQKSSGVKSTHQAAIVSKIDSRLEQYTSAIEGTKSAKPTKPAASDLPVPAEGVRNIKSMWEKGNVFSSPTAAGTPNKETAGLKVGVSSRINEWLTKTPDGNKSPAPKPSDLRPGDVSSKRNLWEKQSVDKVTS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALD1(800)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Caldesmon recombinant protein epitope signature tag (PrEST)

Application

Anti-CALD1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Caldesmon 1 is a cytoskeleton-associated protein encoded by the gene CALD1. It is capable of binding to actin and functions in the stabilization of actin filaments. It can also bind to calmodulin and play a role in the regulation of smooth muscle and nonmuscle contraction. The binding domain of this protein also binds to tropomyosin, myosin, and phospholipids. CALD1 has been implicated in actin cytoskeletal modeling and cell migration. It is associated with actin stress fibers and membrane ruffles, where it plays a role in the polymerization of these fibers. It also functions in cell division, adhesion and apoptosis. It serves as a marker for distinguishing muscularis propria from desmoplasia, facilitating proper staging of colonic adenocarcinoma. It plays a role in gastric cancer metastasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71214

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jordan A Roberts et al.
International journal of clinical and experimental pathology, 7(2), 792-796 (2014-02-20)
An accurate distinction between deep muscularis propria invasion versus subserosal invasion by colonic adenocarcinoma is essential for the accurate staging of cancer and subsequent optimal patient management. However, problems may arise in pathologic staging when extensive desmoplasia blurs the junction
Qian Hou et al.
Journal of proteome research, 12(2), 980-990 (2012-12-26)
In this study, we aim to identify biomarkers for gastric cancer metastasis using a quantitative proteomics approach. The proteins extracted from a panel of 4 gastric cancer cell lines, two derived from primary cancer (AGS, FU97) and two from lymph
Daniele V F Tauriello et al.
Nature, 554(7693), 538-543 (2018-02-15)
Most patients with colorectal cancer die as a result of the disease spreading to other organs. However, no prevalent mutations have been associated with metastatic colorectal cancers. Instead, particular features of the tumour microenvironment, such as lack of T-cell infiltration
Rene Jackstadt et al.
Cancer cell, 36(3), 319-336 (2019-09-19)
The metastatic process of colorectal cancer (CRC) is not fully understood and effective therapies are lacking. We show that activation of NOTCH1 signaling in the murine intestinal epithelium leads to highly penetrant metastasis (100% metastasis; with >80% liver metastases) in
Christoph N Köhler
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 59(5), 504-517 (2011-03-18)
The author has recently reported the distribution of the cytoskeleton-associated protein caldesmon in spleen and lymph nodes detected with different antibodies against caldesmon (J Histochem Cytochem 58:183-193, 2010). Here the author reports the distribution of caldesmon in the CNS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service