Skip to Content
Merck
All Photos(6)

Key Documents

HPA007888

Sigma-Aldrich

Anti-CECR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cat eye syndrome critical region protein 1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TDWQGTSIDRNILDALMLNTTRIGHGFALSKHPAVRTYSWKKDIPIEVCPISNQVLKLVSDLRNHPVATLMATGHPMVISSDDPAMFGAKGLSYDFYEVFMGIGGMKADLRTLKQLAMNSIKYSTLLESEKNTFMEIWKKRWDK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CECR1(51816)

General description

CECR1 (cat eye syndrome chromosome region, candidate 1) gene encodes a member of the family of adenosine deaminase growth factors (ADGFs) that have sequence similarity with classical adenosine deaminase (ADA) at the C-terminus. CECR1 is mainly expressed in fetal heart and kidney and in adult heart, lung and kidney. It contains an ADA (adenosine deaminase) domain and functions by regulating the level of extra-cellular adenosine. It is extensively glycosylated and contains a conserved disulfide bond and a signal peptide. This gene is mapped to human chromosome 22q11.2.

Immunogen

Cat eye syndrome critical region protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CECR1 (cat eye syndrome chromosome region, candidate 1) is also called as ADA2 (adenosine deaminase 2) and plays a role in the degradation of extracellular adenosine, which acts as a signaling molecule in several cellular processes. ADA2 is secreted by differentiating monocytes and may function in the regulation of cell proliferation and differentiation. It is bound to the cell surface via proteoglycans and certain adenosine deaminase receptors. It promotes T cell-dependent differentiation of monocytes into macrophages and promotes proliferation of macrophages. It may serve as a drug candidate to regulate the immune responses during inflammation and cancer. Defects in this gene are associated with Cat eye syndrome and is responsible for some of the phenotype observed in this syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73541

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changbin Zhu et al.
Oncotarget, 9(71), 33500-33514 (2018-10-17)
Tumor associated macrophages (TAMs) promote tumor development, angiogenesis and distal metastasis. In previous studies, we showed that Cat Eye Syndrome Critical Region Protein 1 (CECR1) is expressed by M2-like TAMs in human glioma samples. CECR1 promoted M2 TAMs differentiation and
Andrey V Zavialov et al.
Journal of leukocyte biology, 88(2), 279-290 (2010-05-11)
ADAs play a pivotal role in regulating the level of adenosine, a signaling molecule controlling a variety of cellular responses by binding to and activating four ADRs. Two enzymes, ADA1 and ADA2, are known to possess ADA activity in humans.
Johanna Schepp et al.
Arthritis & rheumatology (Hoboken, N.J.), 69(8), 1689-1700 (2017-05-12)
We aimed to test the relevance of deficiency of adenosine deaminase 2 (DADA2) in patients with antibody deficiency and describe the clinical picture of the disease in adulthood. We screened for DADA2 in a cohort of 181 patients with antibody
M A Riazi et al.
Genomics, 64(3), 277-285 (2000-04-11)
Cat eye syndrome (CES) is a developmental disorder with multiple organ involvement, associated with the duplication of a 2-Mb region of 22q11.2. Using exon trapping and genomic sequence analysis, we have isolated and characterized a gene, CECR1, that maps to
Anton V Zavialov et al.
The Journal of biological chemistry, 285(16), 12367-12377 (2010-02-12)
Two distinct adenosine deaminases, ADA1 and ADA2, are found in humans. ADA1 has an important role in lymphocyte function and inherited mutations in ADA1 result in severe combined immunodeficiency. The recently isolated ADA2 belongs to the novel family of adenosine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service