Skip to Content
Merck
All Photos(6)

Key Documents

HPA007649

Sigma-Aldrich

Anti-MAP1LC3A antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, rabbit polyclonal

Synonym(s):

Anti-Autophagy-related protein LC3 A, Anti-MAP1 light chain 3- like protein 1, Anti-MAP1A/1B light chain 3 A, Anti-MAP1A/MAP1B LC3 A, Anti-Microtubule-associated protein 1 light chain 3 alpha, Anti-Microtubule-associated proteins 1A/1B light chain 3A precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

Product Name

Anti-MAP1LC3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MPSDRPFKQRRSFADRCKEVQQIRDQHPSKIPVIIERYKGEKQLPVLDKTKFLVPDHVNMS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Microtubule-associated proteins 1A/1B light chain 3A precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-MAP1LC3A antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MAP1LC3A (microtubule-associated protein 1 light chain 3 α) gene encodes a light chain subunit that can associate with either MAP1A or MAP1B, which are microtubule-associated proteins involved in the interaction between microtubules and cytoskeleton components. The light chain is an essential component of the autophagic vacuole and serves as a maker of autophagy induction. Its accumulation in colorectal tumor cells serves as a good prognostic marker. MAP1LC3A may play a role in the development of breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71246

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Giatromanolaki et al.
Journal of clinical pathology, 63(10), 867-872 (2010-09-30)
The microtubule-associated protein 1 light chain 3 (LC3A) is an essential component of the autophagic vacuoles, forming a reliable marker of autophagic activity. In a previous study, the authors showed that LC3A immunohistochemistry renders three patterns of autophagic expression in
Youn Ju Lee et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 15(1), 1-7 (2011-04-05)
Many anticancer agents as well as ionizing radiation have been shown to induce autophagy which is originally described as a protein recycling process and recently reported to play a crucial role in various disorders. In HCT116 human colon cancer cells
Ekhlas Qaid Gazem Othman et al.
Journal of clinical laboratory analysis, 23(4), 249-258 (2009-07-23)
Autophagy is a protein degradation process within the cell and its deregulation has been linked to various diseases and the formation of cancer. One of the important proteins involved in the autophagy process is microtubule-associated protein 1 light chain 3
K E Funk et al.
Neuropathology and applied neurobiology, 37(3), 295-306 (2010-10-16)
Granulovacuolar degeneration involves the accumulation of large, double membrane-bound bodies within certain neurones during the course of Alzheimer's disease (AD) and other adult-onset dementias. Because of the two-layer membrane morphology, it has been proposed that the bodies are related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service