Skip to Content
Merck
All Photos(3)

Key Documents

HPA006531

Sigma-Aldrich

Anti-MPG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-3-alkyladenine DNA glycosylase, Anti-3-methyladenine DNA glycosidase, Anti-ADPG, Anti-DNA-3-methyladenine glycosylase, Anti-N-methylpurine-DNA glycosylase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YFCMNISSQGDGACVLLRALEPLEGLETMRQLRSTLRKGTASRVLKDRELCSGPSKLCQALAINKSFDQRDLAQDEAVWLERGPLEPSEPAVVAAARVGVGHAGEWARKPLRFYVRGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPG(4350)

General description

N-methylpurine-DNA glycosylase (MPG) is a mitochondrial DNA repair enzyme. It is expressed in the nucleus and cytoplasm along with mitochondria.

Immunogen

DNA-3-methyladenine glycosylase recombinant protein epitope signature tag (PrEST)

Application

Anti-MPG antibody is suitable for Far-western blotting. Anti-MPG antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

N-methylpurine-DNA glycosylase (MPG) directly interacts with mitochondrial single-stranded binding protein (mtSSB) during mitochondrial DNA repair. The elevated expression has been reported in several tumor cells such as breast, lung, and colon cancers. Its N terminal domain interacts with the DNA binding site of the tumor suppressor gene p53 to represses its activity directly. By interacting with p53, it regulates the p53-mediated cell cycle arrest.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71058

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yael Leitner-Dagan et al.
Journal of the National Cancer Institute, 104(22), 1765-1769 (2012-10-30)
Only a minority of smokers develop lung cancer, possibly due to genetic predisposition, including DNA repair deficiencies. To examine whether inter-individual variations in DNA repair activity of N-methylpurine DNA glycosylase (MPG) are associated with lung cancer, we conducted a blinded
Eleanor Healing et al.
Nucleic acids research, 47(11), e61-e61 (2019-03-15)
DNA repair is essential for the maintenance of genomic integrity, and evidence suggest that inter-individual variation in DNA repair efficiency may contribute to disease risk. However, robust assays suitable for quantitative determination of DNA repair capacity in large cohort and
Natalia M Leguisamo et al.
Oncotarget, 8(33), 54199-54214 (2017-09-15)
Colorectal cancer (CRC) is prevalent worldwide, and treatment often involves surgery and genotoxic chemotherapy. DNA repair mechanisms, such as base excision repair (BER) and mismatch repair (MMR), may not only influence tumour characteristics and prognosis but also dictate chemotherapy response.
Shanshan Song et al.
Cell research, 22(8), 1285-1303 (2012-07-18)
Alkylating agents induce genome-wide base damage, which is repaired mainly by N-methylpurine DNA glycosylase (MPG). An elevated expression of MPG in certain types of tumor cells confers higher sensitivity to alkylation agents because MPG-induced apurinic/apyrimidic (AP) sites trigger more strand
Barbara van Loon et al.
DNA repair, 12(3), 177-187 (2013-01-08)
Due to a harsh environment mitochondrial genomes accumulate high levels of DNA damage, in particular oxidation, hydrolytic deamination, and alkylation adducts. While repair of alkylated bases in nuclear DNA has been explored in detail, much less is known about the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service