Skip to Content
Merck
All Photos(6)

Key Documents

HPA005728

Sigma-Aldrich

Anti-HIVEP3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ16752, Anti-KBP-1, Anti-KBP1, Anti-KIAA1555, Anti-KRC, Anti-SHN3, Anti-Schnurri-3, Anti-ZAS3, Anti-ZNF40C, Anti-human immunodeficiency virus type I enhancer binding protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunofluorescence: 0.25-2 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

SYSFDDHITDSEALSRSSHVFTSHPRMLKRQPAIELPLGGEYSSEEPGPSSKDTASKPSDEVEPKESELTKKTKKGLKTKGVIYECNICGARYKKRDNYEAHKKYYCSELQIAKPISAGTHTSPEAEKSQIEHEPWSQMMHYKLGTTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIVEP3(59269)

Immunogen

human immunodeficiency virus type I enhancer binding protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Human immunodeficiency virus type I enhancer binding protein 3 (HIVEP3) is a protein encoded by the HIVEP3 gene in humans. It is referred as KRC, KBP1, SHN3, ZAS3, KBP-1, ZNF40C and Schnurri-3. The gene is a large zinc finger transcription repressor that binds the κB-motif through two signature domains of ZASN and ZASC. Lack of this protein induces accelerated cell proliferation and tumorigenesis. It represses NFκB-activated transcription by competing with NFκB for the κB-motif. It is a tumor suppressor gene and may serves as a novel therapeutic target for developing anti-cancer drugs. Its locus encodes a signaling and transcriptional molecule involved in regulating inflammatory responses. Its expression is directly regulated by estrogen and is overexpressed in lupus. Up-regulation of this gene by estrogen may play a critical role in female-biased autoimmune disorders. It possess therapeutic potential for controlling aberrant activation of NFκB in various diseases.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86946

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joung-Woo Hong et al.
BMB reports, 43(12), 807-812 (2010-12-30)
NFκB and ZAS3 are transcription factors that control important cellular processes including immunity, cell survival and apoptosis. Although both proteins bind the κB-motif, they produce opposite physiological consequences; NFκB activates transcription, promotes cell growth and is often found to be
Rubén A Bartolomé et al.
Cell death & disease, 14(11), 742-742 (2023-11-15)
Interleukin 13 receptor alpha 2 (IL13Rα2) is a relevant therapeutic target in glioblastoma (GBM) and other tumors associated with tumor growth and invasion. In a previous study, we demonstrated that protein tyrosine phosphatase 1B (PTP1B) is a key mediator of
Dong-Hyeon Shin et al.
BMB reports, 44(4), 267-272 (2011-04-29)
ZAS3 is a large zinc finger transcription repressor that binds the ϰB-motif via two signature domains of ZASN and ZASC. A loss-of-function study showed that lack of ZAS3 protein induced accelerated cell proliferation and tumorigenesis. Conversely, gain-of-function studies showed that
Nicholas A Young et al.
Molecular immunology, 54(1), 23-31 (2012-11-28)
Systemic lupus erythematosus (SLE) is a prototypic, inflammatory autoimmune disease characterized by significant gender bias. Previous studies have established a role for hormones in SLE pathogenesis, including the sex hormone estrogen. Estrogen regulates gene expression by translocating estrogen receptors (ER)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service