Skip to Content
Merck
All Photos(4)

Key Documents

HPA005716

Sigma-Aldrich

Anti-SDC4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Amphiglycan antibody produced in rabbit, Anti-Ryudocan core protein antibody produced in rabbit, Anti-SYND4 antibody produced in rabbit, Anti-Syndecan-4 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

PQDLLEGRYFSGALPDDEDVVGPGQESDDFELSGSGDLDDLEDSMIGPEVVHPLVPLDNHIPERAGSGSQVPTEPKKLEENEVIPKRISPVEESEDVSNKVSMSSTVQGSNIFERTEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SDC4(6385)

General description

Syndecan 4 (SDC4) is a cell surface proteoglycan which binds heparan sulphate. It has membrane-spanning domains, intracellular domains and structurally distinct extracellular domains.

Immunogen

Syndecan-4 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Syndecan 4 (SDC4) acts as a cell surface receptor during cell-cell and cell-matrix interactions. It binds and modifies the action of various growth factors, cytokines, proteases, cell adhesion molecules and extracellular matrix components. SDC4 forms a compact intertwined dimer, stabilized by ionic interactions between peptides at physiological pH with a clamp shape in the central variable V region. It also induces the growth, migration and invasion of hepatoma cells. The association of stromal cell-derived factor 1 (SDF-1), chemokine receptor 4 (CXCR4) and SDC4 induces the growth of human hemochromatotic cell line (Huh7) cells by promoting their entry into the cell cycle. This also inhibits the tumor necrosis factor-α-mediated apoptosis of the cells. During cartilage breakdown in osteoarthritis, it controls the activation of ADAM metallopeptidase with thrombospondin type 1 motif 5 (ADAMTS-5) through direct interaction with the protease and by regulating mitogen-activated protein kinase (MAPK)-dependent synthesis of matrix metalloproteinase-3 (MMP-3).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85154

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syndecan family of cell surface proteoglycans: developmentally regulated receptors for extracellular effector molecules.
M Salmivirta and M Jalkanen
Experientia, 51(9-10) , 863-872 (1995)
S V Subramanian et al.
The Journal of biological chemistry, 272(23), 14713-14720 (1997-06-06)
The syndecan family of transmembrane heparan sulfate proteoglycans is abundant on the surface of all adherent mammalian cells. Syndecans bind and modify the action of various growth factors/cytokines, proteases/antiproteases, cell adhesion molecules, and extracellular matrix components. Syndecan expression is highly
Angela Sutton et al.
Molecular cancer research : MCR, 5(1), 21-33 (2007-01-30)
In addition to their physiologic effects in inflammation and angiogenesis, chemokines are involved in cancer pathology. The aim of this study was to determine whether the chemokine stromal cell-derived factor 1 (SDF-1) induces the growth, migration, and invasion of human
Frank Echtermeyer et al.
Nature medicine, 15(9), 1072-1076 (2009-08-18)
Aggrecan cleavage by a disintegrin and metalloproteinase with a thrombospondin type 1 motif, member 5 (ADAMTS-5) is crucial for the breakdown of cartilage matrix during osteoarthritis, a degenerative joint disease that leads to the progressive destruction of articular structures. The
J Shin et al.
Biochemistry, 40(29), 8471-8478 (2001-07-18)
The syndecans, transmembrane proteoglycans which are involved in the organization of cytoskeleton and/or actin microfilaments, have important roles as cell surface receptors during cell-cell and/or cell-matrix interactions. Since previous studies indicate that the function of the syndecan-4 cytoplasmic domain is

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service