Skip to Content
Merck
All Photos(2)

Key Documents

HPA003747

Sigma-Aldrich

Anti-TRIOBP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DFNB28, Anti-HRIHFB2122, Anti-KIAA1662, Anti-TAP68, Anti-Tara

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EIGALMRQAEEREHTLRRCQQEGQELLRHNQELHGRLSEEIDQLRGFIASQGMGNGCGRSNERSSCELEVLLRVKENELQYLKKEVQCLRDELQMMQKDKRFTSGKYQDVYVELSHIKTRSEREIEQLKEHLRL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIOBP(11078)

Immunogen

TRIO and F-actin-binding protein recombinant protein epitope signature tag (PrEST)

Application

Anti-TRIOBP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

TRIO and F-actin-binding protein is a protein encoded by the TRIOBP gene in humans and is mapped to chromosome 22q13. It is identified as a binding protein of guanine nucleotide exchange factor Trio. It may be associated with many fundamental cellular processes, ranging from actin remodeling, directed cell movement, to cell cycle regulation, aging and cancer. The protein is associated with telomeric repeat binding factor 1 (TRF1) and mediates the function of TRF1 in mitotic regulation. It is a novel Polo-like kinase 1 (Plk1) target protein. Plk1 in cooperation with this gene is important for chromosome segregation during mitotic progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73552

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxiao Li et al.
Protein expression and purification, 55(1), 84-92 (2007-07-17)
Tara was originally identified as a binding protein of guanine nucleotide exchange factor Trio. Although Tara may be involved in many fundamental cellular processes, ranging from actin remodeling, directed cell movement, to cell cycle regulation, aging, and cancer, the exact
Yuanyuan Zhu et al.
Experimental cell research, 318(18), 2344-2352 (2012-07-24)
Trio-associated repeat on actin (Tara) is an F-actin binding protein and regulates actin cytoskeletal organization. In our previous study, we have found that Tara associates with telomeric repeat binding factor 1 (TRF1) and mediates the function of TRF1 in mitotic
Saima Riazuddin et al.
American journal of human genetics, 78(1), 137-143 (2005-12-31)
In seven families, six different mutant alleles of TRIOBP on chromosome 22q13 cosegregate with autosomal recessive nonsyndromic deafness. These alleles include four nonsense (Q297X, R788X, R1068X, and R1117X) and two frameshift (D1069fsX1082 and R1078fsX1083) mutations, all located in exon 6
Nicholas J Bradshaw et al.
The Journal of biological chemistry, 292(23), 9583-9598 (2017-04-26)
Aggregation of specific proteins in the brains of patients with chronic mental illness as a result of disruptions in proteostasis is an emerging theme in the study of schizophrenia in particular. Proteins including DISC1 (disrupted in schizophrenia 1) and dysbindin-1B
Nicholas J Bradshaw et al.
PloS one, 9(10), e111196-e111196 (2014-10-22)
We have previously proposed that specific proteins may form insoluble aggregates as a response to an illness-specific proteostatic dysbalance in a subset of brains from individuals with mental illness, as is the case for other chronic brain conditions. So far

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service