Skip to Content
Merck
All Photos(2)

Key Documents

HPA003128

Sigma-Aldrich

Anti-EP300 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-E1A-associated protein p300, Anti-HAT p300, Anti-Histone acetyltransferase p300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FLPQTQFPSQGMNVTNIPLAPSSGQAPVSQAQMSSSSCPVNSPIMPPGSQGSHIHCPQLPQPALHQNSPSPVPSRTPTPHHTPPSIGAQQPPATTIPAPVPTPPAMPPGPQSQALHPPPRQTPTPPTTQLPQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EP300(2033)

Looking for similar products? Visit Product Comparison Guide

General description

The gene EP300 (E1A binding protein p300) is mapped to human chromosome 22q13.2. The encoded protein contains a HAT domain and binding sites for transcription factors.

Immunogen

Histone acetyltransferase p300 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

EP300 (E1A binding protein p300) gene encodes the transcriptional co-activator protein called p300. It is a histone acetyl transferase that regulates gene expression through various signal-transduction pathways. It is involved in chromatin remodeling, cell proliferation and differentiation. It specifically binds to phosphorylated CREB protein and controls cAMP-gene regulation. In response to DNA damage, it acetylates TP53 and regulates its DNA-binding and transcription functions. It functions as a tumor suppressor. Mutations in this gene may be linked to human cancers. Defects in this gene cause Rubinstein-Taybi syndrome that is characterized by growth retardation, facial dysmorphisms, skeletal abnormalities and mental retardation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73567

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zimam Mahmud et al.
Biochemical pharmacology, 163, 391-403 (2019-03-14)
E-cadherin transcriptional activator EP300 is down-regulated in metaplastic breast carcinoma, a rare form of triple negative and E-cadherin-negative aggressive breast cancer with a poor clinical outcome. In order to shed light on the regulation of E-cadherin by EP300 in breast
Emma J Bryan et al.
International journal of cancer, 102(2), 137-141 (2002-10-18)
The putative tumour suppressor gene EP300 is located on chromosome 22q13 which is a region showing frequent loss of heterozygosity (LOH) in colon, breast and ovarian cancers. We analysed 203 human breast, colon and ovarian primary tumours and cell lines
Deborah Bartholdi et al.
Journal of medical genetics, 44(5), 327-333 (2007-01-16)
Rubinstein-Taybi syndrome (RSTS) is a congenital disorder characterised by growth retardation, facial dysmorphisms, skeletal abnormalities and mental retardation. Broad thumbs and halluces are the hallmarks of the syndrome. RSTS is associated with chromosomal rearrangements and mutations in the CREB-binding protein
Soeren Torge Mees et al.
International journal of cancer, 126(1), 114-124 (2009-07-02)
Genetic and epigenetic alterations during development of pancreatic ductal adenocarcinomas (PDACs) are well known. This study investigates genetic and epigenetic data together with tumor biology to find specific alterations responsible for metastasis formation. Using 16 human PDAC cell lines in
Bo Ram Kim et al.
Molecular & cellular proteomics : MCP, 16(10), 1864-1888 (2017-08-11)
Lung cancer is the leading cause of cancer mortality worldwide, with squamous cell carcinoma (SQCC) being the second most common form. SQCCs are thought to originate in bronchial basal cells through an injury response to smoking, which results in this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service