Skip to Content
Merck
All Photos(10)

Key Documents

HPA002877

Sigma-Aldrich

Anti-PGRMC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Membrane-associated progesterone receptor component 1 antibody produced in rabbit, Anti-mPR antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DQPAASGDSDDDEPPPLPRLKRRDFTPAELRRFDGVQDPRILMAINGKVFDVTKGRKFYGPEGPYGVFAGRDASR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGRMC1(10857)

General description

Progesterone receptor membrane component 1/Sigma-2 receptor (PGRMC1) is a heme-binding protein involved in the activation of P450 proteins. PGRMC1 also binds plasminogen activator inhibitor RNA-binding protein-1 (PAIR-BP1). PGRMC1 is involved in the regulation of cancer cell survival or death following chemotherapy. PGRMC1-regulated gene expression is involved in progesterone inhibition of apoptosis.
Rabbit polyclonal anti-PGRMC1 antibody reacts with human membrane-associated progesterone receptor component 1.

Immunogen

Membrane-associated progesterone receptor component 1 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Western Blotting (1 paper)
Rabbit polyclonal anti-PGRMC1 antibody is used to tag membrane-associated progesterone receptor component 1 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of membrane-associated progesterone receptor component 1 in the regulation of cell survival and apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74350

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Smith et al.
Kidney & blood pressure research, 45(2), 233-248 (2020-02-18)
Diabetic nephropathy (DN) and hypertensive nephrosclerosis (HN) represent the most common causes of chronic kidney disease (CKD) and many patients progress to -end-stage renal disease. Patients are treated primarily through the management of cardiovas-cular risk factors and hypertension; however patients
Valentina Lodde et al.
Cancers, 14(23) (2022-12-12)
During mitosis, chromosome missegregation and cytokinesis defects have been recognized as hallmarks of cancer cells. Cytoskeletal elements composing the spindle and the contractile ring and their associated proteins play crucial roles in the faithful progression of mitotic cell division. The
Terrence K Allen et al.
Reproductive sciences (Thousand Oaks, Calif.), 21(2), 260-268 (2013-07-03)
Progesterone (P4) and the progestin, 17α-hydroxyprogesterone caproate, are clinically used to prevent preterm births (PTBs); however, their mechanism of action remains unclear. Cytokine-induced matrix metalloproteinase 9 (MMP-9) activity plays a key role in preterm premature rupture of the membranes and
Bashar M Thejer et al.
International journal of molecular sciences, 24(7) (2023-04-14)
Sigma-2 receptor (S2R) is a S2R ligand-binding site historically associated with reportedly 21.5 kDa proteins that have been linked to several diseases, such as cancer, Alzheimer's disease, and schizophrenia. The S2R is highly expressed in various tumors, where it correlates
Wei-Shen Chen et al.
Journal of cutaneous pathology, 40(7), 631-638 (2013-04-06)
Sebaceous carcinoma represents a rare and potentially fatal adnexal malignancy. Poorly-differentiated sebaceous carcinoma consisting of infiltrative basaloid tumor cells with inapparent lipid vesicles can mimic basal cell carcinoma (BCC). Conversely, other epithelial tumors can exhibit clear cell histopathology and mimic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service