Skip to Content
Merck
All Photos(2)

Key Documents

HPA002196

Sigma-Aldrich

Anti-IGFBP7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IGF-binding protein 7 antibody produced in rabbit, Anti-IGFBP-rP1 antibody produced in rabbit, Anti-Insulin-like growth factor-binding protein 7 precursor antibody produced in rabbit, Anti-MAC25 protein antibody produced in rabbit, Anti-PGI2-stimulating factor antibody produced in rabbit, Anti-Prostacyclin-stimulating factor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

EQGPSIVTPPKDIWNVTGAQVYLSCEVIGIPTPVLIWNKVKRGHYGVQRTELLPGDRDNLAIQTRGGPEKHEVTGWVLVSPLSKEDAGEYECHASNSQGQA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGFBP7(3490)

General description

Insulin-like growth factor binding protein 7 (IGFBP7) belongs to the IGFBP family. It is widely distributed in normal tissues but less available in the cancerous cells. IGFBPs possess cysteines at the amino- and carboxyl-terminal regions. It is also expressed in human endometrium.

Immunogen

Insulin-like growth factor-binding protein 7 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-IGFBP7 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

IGFBP7 (insulin like growth factor binding protein 7) serves as a growth-suppressing factor and an IGF (insulin like growth factor)-binding protein. It binds to the IGF peptides to regulate various biological activities of the IGFs. It plays a critical role in the Insulin-like growth factor (IGF) system that modulates steroid hormone actions in the endometrium.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85196

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Gambaro et al.
BMC cancer, 15, 135-135 (2015-04-18)
Insulin-like growth factor binding protein 7 (IGFBP7) has been suggested to act as a tumour suppressor gene in various human cancers, yet its role in epithelial ovarian cancer (EOC) has not yet been investigated. We previously observed that IGFBP7 was
E M Rutanen
Human reproduction (Oxford, England), 15 Suppl 3, 173-181 (2000-10-21)
Insulin-like growth factor (IGF) system is one of the growth factor systems that are believed to modulate steroid hormone actions in the endometrium through autocrine/paracrine mechanisms. IGF-I and IGF-II stimulate proliferation and differentiation, and maintain differentiated cell functions in several
Yingchao Nie et al.
Genetics, 212(4), 1227-1239 (2019-06-20)
Colorectal cancer is a complex disease driven by well-established mutations such as APC and other yet to be identified pathways. The GTPase Rab11 regulates endosomal protein trafficking, and previously we showed that loss of Rab11 caused intestinal inflammation and hyperplasia
Daniele V F Tauriello et al.
Nature, 554(7693), 538-543 (2018-02-15)
Most patients with colorectal cancer die as a result of the disease spreading to other organs. However, no prevalent mutations have been associated with metastatic colorectal cancers. Instead, particular features of the tumour microenvironment, such as lack of T-cell infiltration
Y Oh et al.
The Journal of biological chemistry, 271(48), 30322-30325 (1996-11-29)
The mac25 cDNA was originally cloned from leptomeningial cells and subsequently reisolated through differential display as a sequence preferentially expressed in senescent human mammary epithelial cells. The deduced amino acid sequence of the human mac25 propeptide shares a 20-25% identity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service