Skip to Content
Merck
All Photos(10)

Key Documents

HPA002131

Sigma-Aldrich

Anti-Vinculin Antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, rabbit polyclonal

Synonym(s):

Anti-Metavinculin antibody produced in rabbit, Anti-Vinculin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
rat, human, mouse
citations:
3
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

Product Name

Anti-VCL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

RNAi knockdown
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KMIDERQQELTHQEHRVMLVNSMNTVKELLPVLISAMKIFVTTKNSKNQGIEEALKNRNFTVEKMSAEINEIIRVLQLTSWDEDAWASKDTEAMKRALASIDSKLNQAKGWLRDPSASPGDAGEQAIRQILDEAGKVGELCAGKERREI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)

General description

Vinculin (VCL), a ubiquitously expressed actin-binding cytoskeletal protein, is involved with the cytoplasmic adhesion of cell-cell and cell-extracellular matrix. It is composed of an N-terminal globular head connected via a short polyproline-rich linker sequence to the C-terminal tail portion. An intramolecular association between the head and tail hides many ligand-binding sites in the protein.

Immunogen

Vinculin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VCL (vinculin), a regulator of cell adhesion, is localized at the cytoplasmic region of cell-matrix and cell-cell adhesions to regulate various biological processes, including embryogenesis, wound healing and maintenance of tissue integrity. It has the ability to suppress cell migration and can modulate signaling pathways involved in apoptosis. During cell-cell and cell-extracellular matrix adhesion, VCL binds to the α-actinin, an F-actin bundling protein and talin. Upon binding, it stimulates the assembly of these specialized cellular junctions.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84522

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kris A Demali
Trends in biochemical sciences, 29(11), 565-567 (2004-10-27)
The ability of cells to tightly adhere to one another and to the extracellular matrix is fundamentally important in numerous biological processes, including embryogenesis, wound healing and maintenance of tissue integrity. Vinculin, a protein localized at the cytoplasmic face of
Wolfgang H Ziegler et al.
Trends in cell biology, 16(9), 453-460 (2006-08-09)
Vinculin is a ubiquitously expressed actin-binding protein frequently used as a marker for both cell-cell and cell-extracellular matrix (focal adhesion) adherens-type junctions, but its function has remained elusive. Vinculin is made up of a globular head linked to a tail
P A Weller et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(15), 5667-5671 (1990-08-01)
We have determined the complete sequence of human vinculin, a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Comparison of human and chicken embryo vinculin sequences shows that both proteins contain 1066 amino acids and exhibit a high level of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service