Skip to Content
Merck
All Photos(1)

Key Documents

HPA001560

Sigma-Aldrich

Anti-BCHE antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CHE1, Anti-CHE2, Anti-E1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QQLALQWVQKNIAAFGGNPKSVTLFGESAGAASVSLHLLSPGSHSLFTRAILQSGSFNAPWAVTSLYEARNRTLNLAKLTGCSRENETEIIKCLRNKDPQEILLNEAFVVPYGTPLSVNFGPTVD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCHE(590)

General description

BCHE (butyrylcholinesterase) is a serine hydrolase encoded by the gene BCHE. It is related to acetylcholinesterase and hydrolyzes acetylcholine as well as many other esters. It is predominantly produced in the liver, and is found in plasma, intestinal mucosa and white matter of central nervous system. This gene is localized to human chromosome 3q26, spans 70kb, and is composed of four exons and three introns.

Immunogen

Cholinesterase precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-BCHE antibody produced in rabbit is suitable for global protein profiling to find new molecular biomarkers for common, multifactorial disorders.
Anti-BCHE antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Butyrylcholinesterase is widely distributed in the CNS, indicating its involvement in neural functions. It plays an important role in cholinergic neurotransmission and may have a role in other nervous system functions and in neurodegenerative diseases. Defects in this gene cause a deficiency of butyrylcholinesterase leading to prolonged apnea after the use of certain anesthetic drugs, including the muscle relaxants succinylcholine or mivacurium and other ester local anesthetics.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84516

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Herve Delacour et al.
PloS one, 9(7), e101552-e101552 (2014-07-24)
Butyrylcholinesterase deficiency is characterized by prolonged apnea after the use of muscle relaxants (suxamethonium or mivacurium) in patients who have mutations in the BCHE gene. Here, we report a case of prolonged neuromuscular block after administration of suxamethonium leading to
André Brooking Negrão et al.
PloS one, 8(11), e80505-e80505 (2013-12-07)
The search for genetic vulnerability factors in cocaine dependence has focused on the role that neuroplasticity plays in addiction. However, like many other drugs, the ability of an individual to metabolize cocaine can also influence susceptibility to dependence. Butyrylcholinesterase (BChE)
Ian R Macdonald et al.
Biochemistry, 51(36), 7046-7053 (2012-08-21)
Acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) catalyze the hydrolysis of the neurotransmitter acetylcholine and, thereby, function as coregulators of cholinergic neurotransmission. For both enzymes, hydrolysis takes place near the bottom of a 20 Å deep active site gorge. A number of
C F Bartels et al.
American journal of human genetics, 50(5), 1104-1114 (1992-05-01)
The J-variant of human serum butyrylcholinesterase (BChE) causes both an approximately two-thirds reduction of circulating enzyme molecules and a corresponding decrease in the level of BChE activity present in serum. Since the level of serum BChE activity and the duration
Comparison of butyrylcholinesterase and acetylcholinesterase.
A Chatonnet et al.
The Biochemical journal, 260(3), 625-634 (1989-06-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service