Skip to Content
Merck
All Photos(5)

Key Documents

HPA001473

Sigma-Aldrich

Anti-CASP9 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APAF-3 antibody produced in rabbit, Anti-Apoptotic protease Mch-6 antibody produced in rabbit, Anti-Apoptotic protease-activating factor 3 antibody produced in rabbit, Anti-CASP-9 antibody produced in rabbit, Anti-Caspase-9 precursor antibody produced in rabbit, Anti-ICE-LAP6 antibody produced in rabbit, Anti-ICE-like apoptotic protease 6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ALESLRGNADLAYILSMEPCGHCLIINNVNFCRESGLRTRTGSNIDCEKLRRRFSSLHFMVEVKGDLTAKKMVLALLELAQQDHGALDCCVVVILSHGCQASHLQFPGAVYGTDGCPVSVEKIVNIFNGTSC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP9(842)

General description

CASP9 (Caspase-9) gene encodes a protein belonging to the cysteine-aspartic acid protease (caspase) family. It consists of two domains. The large subunit of caspase 9 possesses a seven-residue loop at the C terminal end. The loop helps to interact across the dimer interface.

Immunogen

Caspase-9 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CASP9 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

CASP9 (Caspase-9) is involved in the apoptotic pathway. The caspases exist as inactive proenzymes that are activated upon proteolytic processing at conserved aspartic residues. The active enzyme is a dimer of one small and one large subunit. It functions as an initiator caspase that is activated by mitochondrial-mediated apoptotic pathway. It executes apoptosis by activating a cascade of effector caspases. It also acts as a tumor suppressor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79896

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Soengas et al.
Science (New York, N.Y.), 284(5411), 156-159 (1999-04-02)
The ability of p53 to promote apoptosis in response to mitogenic oncogenes appears to be critical for its tumor suppressor function. Caspase-9 and its cofactor Apaf-1 were found to be essential downstream components of p53 in Myc-induced apoptosis. Like p53
Michelle M Hill et al.
Molecular interventions, 3(1), 19-26 (2004-03-03)
Apoptosis (programmed cell death) is a physiological process used to eliminate superfluous, damaged, infected, or aged cells in multicellular organisms. During apoptosis the cellular architecture is dismantled from within in a highly controlled fashion. Members of the caspase family of
Anuradha Cingeetham et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(9), 8813-8822 (2014-06-01)
Abnormal apoptosis is one of the hallmarks of cancers including acute myeloid leukemia (AML), as it plays a pivotal role in precisely maintaining self-renewal, proliferation, and differentiation properties of hematopoietic stem cells (HSCs). Caspase9 (CASP9), an initiator caspase activated by
The intrinsic or the extrinsic pathways of apoptosis in the epidermis after cladribine application?
Wrzos PC, et al.
Journal of Biomedical Science and Engineering, 6 (2013)
Michael W Ronellenfitsch et al.
Brain pathology (Zurich, Switzerland), 28(1), 94-102 (2016-12-10)
We report a novel CASP9 germline mutation that may increase susceptibility to the development of brain tumors. We identified this mutation in a family in which three brain tumors had developed within three generations, including two anaplastic astrocytomas occurring in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service