Skip to Content
Merck
All Photos(5)

Key Documents

HPA001471

Sigma-Aldrich

Anti-C1QC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Complement C1q subcomponent subunit C precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

GIPGEPGEEGRYKQKFQSVFTVTRQTHQPPAPNSLIRFNAVLTNPQGDYDTSTGKFTCKVPGLYYFVYHASHTANLCVLLYRSGVKVVTFCGHTSKTNQVNSGGVLLRLQVGEEVWLAVNDYYDMVGIQGSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C1QC(714)

Immunogen

Complement C1q subcomponent subunit C precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

C1QC (complement component 1, q subcomponent, C chain) gene encodes the C-chain polypeptide, a major component of the human complement subcomponent C1q. C1q associates with C1r and C1s proenzymes to form the first component of the serum complement system, C1, which functions in the host defense against pathogens. C1q consists of 18 polypeptide chains comprising six A-chains, six B-chains, and six C-chains. Each of these chains contains a collagen-like region at the N-terminus and a globular region at the C-terminus. These chains are localized to chromosome1 and are present as A-C-B. Deficiency of C1q is linked to lupus erythematosus and glomerulonephritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78219

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Berkel et al.
European journal of pediatrics, 156(2), 113-115 (1997-02-01)
A 7-year-old male with recurrent erythematous and desquamated skin lesions and respiratory infections was diagnosed as selective complete C1q deficiency following detailed studies of the complement system. His asymptomatic sister also had selective complete C1q deficiency. During a follow up
Proteins involved in the activation and control of the two pathways of human complement.
K B Reid
Biochemical Society transactions, 11(1), 1-12 (1983-01-01)
Berhane Ghebrehiwet et al.
Current directions in autoimmunity, 7, 87-97 (2004-01-15)
The association between C1q and autoimmune diseases such as rheumatoid arthritis and systemic lupus erythematosus (SLE) is well established. Deficiency in C1q is considered to be a strong susceptibility factor and is corroborated by the fact that > or =
Structure and activation of the C1 complex of complement: unraveling the puzzle.
Christine Gaboriaud et al.
Trends in immunology, 25(7), 368-373 (2004-06-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service