Skip to Content
Merck
All Photos(1)

Key Documents

HPA000886

Sigma-Aldrich

Anti-FRMD7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FERM domain-containing protein 7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

QVPRWSPIRAEERTSPHSYVEPTAMKPAERSPRNIRMKSFQQDLQVLQEAIARTSGRSNINVGLEEEDPNLEDAFVCNIQEQTPKRSQSQSDMKTIRFPFGSEFRPLGPCPALSHKADLFTDMFAEQELPAVLMDQSTAERYVASESSDS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FRMD7(90167)

General description

The gene FRMD7 (FERM domain containing 7) is mapped to human chromosome Xq27. The encoded FERM-domain containing protein is found to be expressed in the ventricular layer of the forebrain, midbrain, cerebellar primordium, spinal cord and the developing neural retina of human embryo.

Immunogen

FERM domain-containing protein 7 recombinant protein epitope signature tag (PrEST)

Application

Anti-FRMD7 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

FERM domain-containing protein 7 is a protein encoded by the FRMD7 gene in humans. Restricted expression of FRMD7 in human is found in embryonic brain and developing neural retina and may have a role in the control of eye movement and gaze stability. The gene is linked to Idiopathic congenital nystagmus (ICN), which is a genetically heterogeneous disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74162

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-wei Song et al.
Journal of Zhejiang University. Science. B, 14(6), 479-486 (2013-06-05)
To screen mutations in FERM domain-containing protein 7 (FRMD7) gene in two Chinese families with X-linked idiopathic congenital nystagmus (XLICN). Common ophthalmic data and peripheral blood of two Chinese XLICN families (families A and B) were collected after informed consent.
Zhirong Liu et al.
Molecular vision, 19, 1834-1840 (2013-08-16)
Idiopathic congenital nystagmus (ICN) is a genetically heterogeneous disease. Thus far, the disease gene has been identified as the FERM domain containing 7 (FRMD7) gene. The purpose of this study was to elucidate the clinical and genetic characteristics of a
Jiali Pu et al.
Molecular vision, 17, 591-597 (2011-03-10)
FERM domain containing 7 (FRMD7) is a member of the four-point-one, ezrin, radixin, moesin (FERM) family of proteins, and has been reported to cause X-linked idiopathic congenital nystagmus (ICN), a disease which affects ocular motor control. There have been over
Patrick Tarpey et al.
Nature genetics, 38(11), 1242-1244 (2006-10-03)
Idiopathic congenital nystagmus is characterized by involuntary, periodic, predominantly horizontal oscillations of both eyes. We identified 22 mutations in FRMD7 in 26 families with X-linked idiopathic congenital nystagmus. Screening of 42 singleton cases of idiopathic congenital nystagmus (28 male, 14
Daniel F Schorderet et al.
Human mutation, 28(5), 525-525 (2007-04-03)
Congenital nystagmus is an eye movement disorder in which one or both eyes are in constant movement. It can be associated with a number of ocular or neurological diseases, or it can be inherited in an autosomal or X-linked fashion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service