Skip to Content
Merck
All Photos(9)

Key Documents

HPA000835

Sigma-Aldrich

Anti-NPC2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Epididymal secretory protein E1 precursor antibody produced in rabbit, Anti-Niemann-Pick disease type C2 protein antibody produced in rabbit, Anti-hE1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

PVQFKDCGSVDGVIKEVNVSPCPTQPCQLSKGQSYSVNVTFTSNIQSKSSKAVVHGILMGVPVPFPIPEPDGCKSGINCPIQKDKTYSYLNKLPVKSEYPSIKLVVEWQLQDDKNQSLFCWEIPVQIVSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPC2(10577)

General description

Niemann Pick type C (NPC2) protein is a member of the Niemann Pick type C family of proteins.
The gene NPC2 (Niemann-Pick disease type C2) is mapped to human chromosome 14q24.3. It contains only five exons spanning a length of 13.5kb and encodes a soluble lysosomal protein of 132 amino acids.

Immunogen

Epididymal secretory protein E1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-NPC2 antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

Niemann-Pick type C is a lysosomal storage disease (fatal autosomal recessive disorder) caused due to mutations in NPC1 and NPC2 genes. It is an inherited disorder in which cholesterol and other lipids accumulate in the late endosomal/lysosomal compartment. The disease is characterized by hepatosplenomegaly and progressive neurological deterioration. NPC2 protein is one of the most abundant components of the epididymal fluid and contains a functional cholesterol-binding site that can transfer cholesterol between membranes. It has a key role for cellular cholesterol regulation in the innate immune response. The protein can be involved in regulation of cholesterol levels in spermatozoa during epididymal maturation. NPC2 acts as a specific regulator of arachidonic acid (AA) metabolism and inflammation and may help in the treatment of inflammatory diseases characterized by the presence of activated fibroblasts.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70511

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milan Hodošček et al.
International journal of molecular sciences, 19(9) (2018-09-06)
The Niemann Pick type C (NPC) proteins, NPC1 and NPC2, are involved in the lysosomal storage disease, NPC disease. The formation of a NPC1⁻NPC2 protein⁻protein complex is believed to be necessary for the transfer of cholesterol and lipids out of
Andrey Frolov et al.
The Journal of biological chemistry, 288(33), 23696-23703 (2013-07-03)
Activated fibroblasts, also known as myofibroblasts, are mediators of several major human pathologies including proliferative fibrotic disorders, invasive tumor growth, rheumatoid arthritis, and atherosclerosis. We previously identified Niemann-Pick type C2 (NPC2) protein as a negative regulator of fibroblast activation (Csepeggi
Blair R Roszell et al.
PloS one, 8(7), e67084-e67084 (2013-07-12)
Niemann-Pick C (NPC) disease is due to loss of NPC1 or NPC2 protein function that is required for unesterified cholesterol transport from the endosomal/lysosomal compartment. Though lung involvement is a recognized characteristic of Niemann-Pick type C disease, the pathological features
Afagh Alavi et al.
Molecular genetics and metabolism, 110(1-2), 139-144 (2013-06-25)
We report identification of a homozygous mutation in NPC2 in two Iranian siblings with a neurologic dysfunction whose disease had not been diagnosed prior to our genetic analysis. The mutation was identified by exome sequencing. The finding resulted in diagnosis
Heiko Runz et al.
Human mutation, 29(3), 345-350 (2007-12-18)
Niemann-Pick type C (NPC) disease is a rare autosomal-recessive lysosomal storage disease typically accompanied by progressive impairment of nervous system and liver function. Biochemically, the disorder presents with an inhibited egress of cholesterol and glycosphingolipids from endosomal and lysosomal compartments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service