Skip to Content
Merck
All Photos(4)

Key Documents

HPA000657

Sigma-Aldrich

Anti-ACIN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Acinus antibody produced in rabbit, Anti-Apoptotic chromatin condensation inducer in the nucleus antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

QEEPPAKLLDDLFRKTKAAPCIYWLPLTDSQIVQKEAERAERAKEREKRRKEQEEEEQKEREKEAERERNRQLEREKRREHSRERDRERERERERDRGDRDRDRERDRERGRERDRRDTKRHSRSRSRSTPV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACIN1(22985)

General description

ACIN1 (apoptotic chromatin condensation inducer 1) in the nucleus is a protein encoded by the ACIN1 gene in humans. It is also known as Acinus. It exists in three different isoforms termed Acinus-L, Acinus-S′, and Acinus-S. Structurally, all the isoforms are different in their N-terminus while the C-terminus is consistent in all isoforms.

Immunogen

Apoptotic chromatin condensation inducer in the nucleus recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ACIN1 (apoptotic chromatin condensation inducer 1) is associated with several processes such as apoptosis, RNA processing and regulation of gene transcription including RA-dependent transcription. It is essential for the induction of apoptotic chromatin condensation in cells. Hypermethylation of the ACIN1 gene occurs in the early stages of lung adenocarcinoma. It may have a role in the development and malignant transformation of lung adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70439

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Wang et al.
Journal of cellular biochemistry, 115(12), 2165-2174 (2014-08-01)
Acinus has been reported to function in apoptosis, RNA processing and regulation of gene transcription including RA-dependent transcription. There are three different isoforms of Acinus termed Acinus-L, Acinus-S', and Acinus-S. The isoforms of Acinus differ in their N-terminus while the
S Sahara et al.
Nature, 401(6749), 168-173 (1999-09-18)
Apoptosis is defined by several unique morphological nuclear changes, such as chromatin condensation and nuclear fragmentation. These changes are triggered by the activation of a family of cysteine proteases called caspases, and caspase-activated DNase (CAD/DFF40) and lamin protease (caspase-6) have
Yujian Shu et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 1(2), 160-167 (2007-04-06)
In recent years, many studies have performed genome-wide searching for differentially methylated genes in cancer. We hypothesized that characteristic aberrant hypermethylation of CpG islands of certain genes may exist in the early stages of lung adenocarcinoma and that such alterations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service