Skip to Content
Merck
All Photos(7)

Documents

HPA000390

Sigma-Aldrich

Anti-AUTS2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Autism susceptibility gene 2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NSSSSVHPGPLASMPMTVGVTGIHPMNSISSLDRTRMMTPFMGISPLPGGERFPYPSFHWDPIRDPLRDPYRELDIHRRDPLGRDFLLRNDPLHRLSTPRLYEADR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AUTS2(26053)

General description

Autism susceptibility gene 2 (AUTS2) is a part of polycomb repressive complex 2 (PRC2). The gene is located on human chromosome 7q11.22. AUTS2 consists of two proline-rich regions (PR1 and PR2), eight histidine repeats, nuclear localizing sequences and one predicted PY motif. It is expressed in the brain.

Immunogen

Autism susceptibility gene 2 protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-AUTS2 antibody produced in rabbit has been used in:
  • immunohistochemistry
  • immunoprecipitation
  • immunofluorescence
  • western blotting(10)

Biochem/physiol Actions

Autism susceptibility gene 2 (AUTS2) functions as a chromatin modulator, which regulates early lymphoid development and T-cell leukemogenesis. It is associated with autism, alcoholism and heroin dependence. AUTS2 alters Rac signaling. Mutations in this gene is associated with autism spectrum disorders (ASDs), developmental delay and intellectual disability. AUTS2 is implicated in cytoskeleton regulation, transcriptional regulation and neural development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76148

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deregulation of polycomb repressor complex 1 modifier AUTS2 in T-cell leukemia
Nagel S, et al.
Oncotarget, 7(29), 45398-45398 (2016)
A mouse mutation that dysregulates neighboring Galnt17 and Auts2 genes is associated with phenotypes related to the human AUTS2 syndrome
Weisner PA, et al.
G3: Genes, Genomes, Genetics, 29(10), g3-400723 (2019)
Exonic deletions of AUTS2 in Chinese patients with developmental delay and intellectual disability
Fan Y, et al.
American Journal of Medical Genetics. Part A, 170(2), 515-522 (2016)
A genome-wide association study of antidepressant response in Koreans
Myung W, et al.
Translational Psychiatry, 5(9), e633-e633 (2015)
Genome-wide distribution of Auts2 binding localizes with active neurodevelopmental genes
Oksenberg N, et al.
Translational Psychiatry, 4(9), e431-e431 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service