Skip to Content
Merck
All Photos(1)

Key Documents

H8759

Sigma-Aldrich

4-Hydroxy-3-methoxyphenylglycol sulfate potassium salt

≥98% (HPLC), powder

Synonym(s):

(3-Methoxy-4-sulfonyloxyphenyl)glycol potassium salt, MHPG sulfate potassium salt, MOPEG sulfate potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11KO7S
CAS Number:
Molecular Weight:
302.34
Beilstein:
4625651
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

[K+].COc1cc(ccc1OS([O-])(=O)=O)C(O)CO

InChI

1S/C9H12O7S.K/c1-15-9-4-6(7(11)5-10)2-3-8(9)16-17(12,13)14;/h2-4,7,10-11H,5H2,1H3,(H,12,13,14);/q;+1/p-1

InChI key

XFZJWBFZLNLKNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Norepinephrine metabolite that is often used as an index of adrenergic stimulation.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K W Perry et al.
Journal of neural transmission (Vienna, Austria : 1996), 104(8-9), 953-966 (1997-01-01)
The selective serotonin uptake inhibitor fluoxetine (10 mg/kg i.p.) increased tissue levels of the norepinephrine metabolite 3-methoxy-4-hydroxyphenylethylene glycol sulfate (MHPG-SO4) in rat hypothalamus, indicating an increased release of norepinephrine. Microdialysis studies in conscious rats showed that fluoxetine (10 mg/kg i.p.)
G Eisenhofer et al.
Journal of neurochemistry, 66(4), 1565-1573 (1996-04-01)
This study used regional differences in plasma concentrations of norepinephrine and its metabolites to examine how production of the transmitter by sympathetic nerves, in particular, those innervating mesenteric organs, is integrated with metabolism by the liver and elimination by the
E T Hellriegel et al.
Pharmacology, biochemistry, and behavior, 57(1-2), 207-214 (1997-05-01)
The objective of this investigation was to examine the immediate and long term effects of acute, chronic and chronic intermittent stress on the central noradrenergic system of rats. Male Sprague-Dawley rats were subjected to one hour of physical immobilization stress
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service