Skip to Content
Merck
All Photos(1)

Key Documents

H6891

Sigma-Aldrich

7β-Hydroxycholesterol

≥95%

Synonym(s):

5-Cholestene-3β,7β-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95%

form

powder

functional group

hydroxyl

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=C[C@@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h16-18,20-25,28-29H,6-15H2,1-5H3

InChI key

OYXZMSRRJOYLLO-UHFFFAOYSA-N

Application

7β-hydroxycholesterol was used to study oxysterol-induced apoptosis in human endothelial cells.

Biochem/physiol Actions

7β-Hydroxycholesterol is an oxysterol, the enzymatic or non-enzymatic product of cholesterol oxidation. Oxysterols are cytotoxic and induce death in monocytes, smooth muscle cells and endothelial cells. The mechanism of apoptosis induced by oxysterols may involve caspases or DNA fragmentation. Increased levels of 7β-Hydroxycholesterol correlates with increased risk of cardiovascular diseases including atherosclerosis.

Preparation Note

7β-Hydroxycholesterol yields clear, colorless to faint yellow solution in ethanol, with or without heating.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ziedén et al.
Arteriosclerosis, thrombosis, and vascular biology, 19(4), 967-971 (1999-04-09)
The mortality in coronary heart disease among 50- to 54-year-old men is 4 times higher in Lithuania than in Sweden. It was recently suggested that traditional risk factors could not explain this mortality difference. LDL of Lithuanian men showed, however
Ludovic Clarion et al.
Biochemical pharmacology, 83(1), 37-46 (2011-10-11)
7β-Hydroxycholesterol cytotoxicity has been shown in vivo and in vitro to be dependent on the accumulation of its esters. We show in our study, using a detergent-free raft preparation and LC/MS lipid content analysis, that membrane microdomains isolated from 7β-hydroxycholesterol-treated
G Lizard et al.
FEBS letters, 419(2-3), 276-280 (1998-01-15)
The oxysterols, 7beta-hydroxycholesterol and 7-ketocholesterol, are involved in the cytotoxicity of oxidized LDL. To elucidate their molecular mechanisms, the human promonocytic leukemia cells U937 and U4 were used. U4 cells overexpressing Bcl-2 were obtained by transfection of U937 cells. 7Beta-hydroxycholesterol
Raymond C S Seet et al.
Free radical research, 47(4), 283-290 (2013-01-25)
The purpose of this study was to evaluate the use of Framingham risk scores (FRRs) to identify high-risk individuals with biochemical evidence of increased oxidative damage, who may benefit from antioxidant therapies. A bimodal change in plasma F2-isoprostane levels was
Paola Gamba et al.
Aging cell, 10(3), 403-417 (2011-01-29)
All three cholesterol oxidation products implicated thus far in the pathogenesis of Alzheimer's disease, 7β-hydroxycholesterol, 24-hydroxycholesterol, and 27-hydroxycholesterol, markedly enhance the binding of amyloid-beta (Aβ) to human differentiated neuronal cell lines (SK-N-BE and NT-2) by up-regulating net expression and synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service