Skip to Content
Merck
All Photos(1)

Key Documents

H6390

Sigma-Aldrich

Hemin−Agarose

Type I, saline suspension

Synonym(s):

Hemin Agarose, Hemin Binding Gel, Hemin-Agarose Gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

hemin from bovine

Quality Level

type

Type I

form

saline suspension

extent of labeling

≥4 μmol (per ml)

technique(s)

affinity chromatography: suitable

matrix

Cross-linked 4% beaded agarose

matrix activation

epoxy

matrix attachment

carboxyl

matrix spacer

12 atoms

suitability

suitable for chromatography

storage temp.

2-8°C

Application

Hemin Type I-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and specialty resins. Hemin-agarose has been used to evaluate protection against invasion by Gram-negative bacteria.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C S Bracken et al.
Journal of bacteriology, 181(19), 6063-6072 (1999-09-28)
The abilities of two bacterial active heme transporters, HmbR of Neisseria meningitidis and HemR of Yersinia enterocolitica, to use different heme sources were compared. While HmbR-expressing cells used only hemoglobin (Hb) and heme, HemR-expressing bacteria were able to grow on
Swapna Asuthkar et al.
Infection and immunity, 75(9), 4582-4591 (2007-06-20)
In an earlier study, based on the ferric enterobactin receptor FepA of Escherichia coli, we identified and modeled a TonB-dependent outer membrane receptor protein (LB191) from the genome of Leptospira interrogans serovar Lai. Based on in silico analysis, we hypothesized
S Yamamoto et al.
FEMS microbiology letters, 128(2), 195-200 (1995-05-01)
Several clinical isolates of Vibrio parahaemolyticus were examined for their ability to utilize either hemin or hemoglobin as a sole source of iron. Both compounds appeared to be equally good iron sources. Maximum growth was obtained at 5 microM hemin
Inducible glutathione S-transferase (IrGST1) from the tick Ixodes ricinus is a haem-binding protein
Perner J, et al.
Insect Biochemistry and Molecular Biology, 95, 44-54 (2018)
Xueya Liang et al.
Journal of leukocyte biology, 86(2), 229-235 (2009-04-28)
Detection of LPS in tissues is an integral component of innate immunity that acts to protect against invasion by Gram-negative bacteria. Plasma down-regulates LPS-induced cytokine production from macrophages, thereby limiting systemic inflammation in blood and distant tissues. To identify the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service