Skip to Content
Merck
All Photos(1)

Documents

H6166

Sigma-Aldrich

Interferon α 2B human

IFN-α 2B, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

Synonym(s):

IFN α 2B, IFN Alfa 2b, INF α 2B, INF Alfa 2b, Interferon Alfa 2b

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.12 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 16 kDa (glycosylated)

packaging

poly bottle of 10 μg (in poly bottle)

manufacturer/tradename

HumanZyme

technique(s)

cell culture | mammalian: suitable

impurities

≤1EU/mg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IFNA2(3440)

General description

The interferon α 2 (IFN-α2) exists in three forms 2a, 2b and 2c. It consists of 165 amino acids and five α helices. IFN-α2B is found structurally stable at pH 7. The recombinant IFN-α is nonglycosylated compared to the natural human IFN-α.

Biochem/physiol Actions

Interferon α 2B (IFN-α 2B) belongs to the type I interferon protein family. It is produced by many cell types in response to viral infection. IFN-α 2B has antiproliferative and antitumor activities, as well as induction of apoptosis and the inflammatory response. It enhances the expression of MHC proteins by increasing the presentation of antigen peptides and activates cytotoxic T-cells.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine IFN-α 2B is a glycosylated monomer expressed in HEK 293 cells with an apparent molecular mass of 16 kDa. Production in human 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent proliferation of WISH (heteroploid human amnion cell line) in presence of VSV ( vesicular stomatitis virus).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yining Wang et al.
Scientific reports, 11(1), 23465-23465 (2021-12-08)
Human coronavirus NL63 (HCoV-NL63) mainly affects young children and immunocompromised patients, causing morbidity and mortality in a subset of patients. Since no specific treatment is available, this study aims to explore the anti-SARS-CoV-2 agents including favipiravir and remdesivir for treating
Pengfei Li et al.
Science advances, 8(3), eabj5908-eabj5908 (2022-01-20)
Hepatotropic viruses naturally have narrow host and tissue tropisms, challenging the development of robust experimental models. The advent of organoid technology provides a unique opportunity for moving the field forward. Here, we demonstrate that three-dimensional cultured organoids from fetal and
Yingxian Yin et al.
Journal of medical virology, 91(8), 1440-1447 (2019-03-23)
Plaque assay plays an irreplaceable role in a variety of virological studies, including determining titers of viruses. Our previous study showed that a simple and highly repeatable plaque assay could be used for enterovirus 71 (EV-A71). Now, we show that
Yang Li et al.
Antiviral research, 176, 104743-104743 (2020-02-15)
Enteric viruses including hepatitis E virus (HEV), human norovirus (HuNV), and rotavirus are causing global health issues. The host interferon (IFN) response constitutes the first-line defense against viral infections. Melanoma Differentiation-Associated protein 5 (MDA5) is an important cytoplasmic receptor sensing
Formulation and stability of cytokine therapeutics
Lipiainen T, et al.
Journal of Pharmaceutical Sciences, 104(2), 307-326 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service