Skip to Content
Merck
All Photos(2)

Key Documents

H5136

Sigma-Aldrich

Hydromorphone hydrochloride

Synonym(s):

(5α)-4,5-Epoxy-3-hydroxy-17-methyl-morphinan-6-one hydrochloride, Hydromorphone HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3 · HCl
CAS Number:
Molecular Weight:
321.80
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: >10 mg/mL

application(s)

forensics and toxicology
veterinary

SMILES string

OC1=C(O2)C3=C(C[C@@H]4[C@@]5([H])[C@@]3([C@]2([H])C(CC5)=O)CCN4C)C=C1.[H]Cl

InChI

1S/C17H19NO3.ClH/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;/h2,4,10-11,16,19H,3,5-8H2,1H3;1H/t10-,11+,16-,17-;/m0./s1

InChI key

XHILEZUETWRSHC-NRGUFEMZSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Narcotic opiate analgesic; μ opioid receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H M Korani et al.
Veterinary and comparative orthopaedics and traumatology : V.C.O.T, 28(6), 409-416 (2015-10-10)
Evaluate variability associated with assessing changes in the position of uncemented femoral stems. Stem level, canal fill, stem angle, and version angle were measured on craniocaudal horizontal beam (CCHB) and open leg lateral (OLL) radiographic projections of the femur of
E M Costa et al.
Life sciences, 50(1), 73-81 (1992-01-01)
SH-SY5Y (human neuroblastoma) cultured cells, known to have mu-opioid receptors, have been used to assess and compare the ability of eight representative mu-selective compounds from diverse opioid families to recognize and activate these receptors. A wide range of receptor affinities
D L Cichewicz et al.
The Journal of pharmacology and experimental therapeutics, 289(2), 859-867 (1999-04-24)
The antinociceptive effects of various mu opioids given p.o. alone and in combination with Delta-9-tetrahydrocannabinol (Delta9-THC) were evaluated using the tail-flick test. Morphine preceded by Delta9-THC treatment (20 mg/kg) was significantly more potent than morphine alone, with an ED50 shift
P Sacerdote et al.
British journal of pharmacology, 121(4), 834-840 (1997-06-01)
1. Although it is well known that morphine induces significant immunosuppression, the potential immunosuppressive activity of morphine derived drugs commonly used in the treatment of pain (codeine, hydromorphone, oxycodone) has never been evaluated. 2. We evaluated in the mouse the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service