Skip to Content
Merck
All Photos(1)

Key Documents

H3398

Sigma-Aldrich

DL-β-Hydroxylauric acid

≥99% (GC)

Synonym(s):

3-Hydroxydodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O3
CAS Number:
Molecular Weight:
216.32
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (GC)

form

solid

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCC(O)CC(O)=O

InChI

1S/C12H24O3/c1-2-3-4-5-6-7-8-9-11(13)10-12(14)15/h11,13H,2-10H2,1H3,(H,14,15)

InChI key

MUCMKTPAZLSKTL-UHFFFAOYSA-N

Biochem/physiol Actions

DL-3-Hydroxydodecanoic acid (3HDD) is a racemic mixture of D- and L-3HDD. 3-hydroxydodecanoic acid is found as a monomer in the construction of polyhydroxyalkanoates. 3HDD is used to study the roles of β-hydroxy fatty acids in chronic inflammation and insulin resistance and the biosynthesis of diffusible signal factors involved in quorum sensing.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Ping Ouyang et al.
Biomacromolecules, 8(8), 2504-2511 (2007-07-31)
Pseudomonas putida KT2442 produces medium-chain-length (MCL) polyhydroxyalkanoates (PHA) consisting of 3-hydroxyhexanoate (HHx), 3-hydroxyoctanoate (HO), 3-hydroxydecanoate (HD), and 3-hydroxydodecanoate (HDD) from a wide-range of carbon sources. In this study, fadA and fadB genes encoding 3-ketoacyl-CoA thiolase and 3-hydroxyacyl-CoA dehydrogenase in P.
N Ravenscroft et al.
Journal of bacteriology, 174(23), 7595-7605 (1992-12-01)
The lipopolysaccharide (LPS) of the outer membrane of Caulobacter crescentus was purified and analyzed. Two distinct strains of the species, NA 1000 and CB2A, were examined; despite differences in other membrane-related polysaccharides, the two gave similar LPS composition profiles. The
Daniel K Y Solaiman et al.
Biotechnology letters, 28(3), 157-162 (2006-02-21)
Pseudomonas corrugata was selected from a screening process for the bioconversion of inexpensive soy molasses into medium-chain-length poly(hydroxyalkanoates) (mcl-PHA). We obtained yields of 1.5 g cell dry weight (CDW)/l culture with growth medium supplemented with 2% (w/v) soy molasses, and
I MacArthur et al.
Journal of bacteriology, 193(18), 4726-4735 (2011-07-19)
PagL and LpxO are enzymes that modify lipid A. PagL is a 3-O deacylase that removes the primary acyl chain from the 3 position, and LpxO is an oxygenase that 2-hydroxylates specific acyl chains in the lipid A. pagL and
Hongkai Bi et al.
Molecular microbiology, 83(4), 840-855 (2012-01-10)
Signal molecules of the diffusible signal factor (DSF) family have been shown recently to be involved in regulation of pathogenesis and biofilm formation in diverse Gram-negative bacteria. DSF signals are reported to be active not only on their cognate bacteria

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service