Skip to Content
Merck
All Photos(1)

Documents

H0654

Sigma-Aldrich

Hygromycin B solution from Streptomyces hygroscopicus

≥60% (HPLC), 45-60 mg/mL in H2O

Synonym(s):

Hygromycin B, Hygrovetine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
Beilstein:
6755837
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces hygroscopicus

Quality Level

sterility

0.22 μm filtered

concentration

≥60% (HPLC)
45-60 mg/mL in H2O

color

brown-yellow
light orange to dark orange
light yellow to dark yellow

antibiotic activity spectrum

viruses

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CN[C@H]1C[C@@H](N)[C@H](O)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H]3OC4(O[C@H]([C@H](N)CO)[C@H](O)[C@H](O)[C@H]4O)O[C@H]23)[C@@H]1O

InChI

1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6-,7+,8-,9+,10-,11+,12-,13+,14-,15-,16+,17+,18-,19+,20+/m1/s1

InChI key

GRRNUXAQVGOGFE-XKIAHZFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Hygromycin B is an aminoglycoside antibiotic isolated from Streptomyces hygroscopicus. It has been used to study protein synthesis at the level of the 70S ribosome translocation and mRNA template misreading, as an antiviral agent by selectively penetrating cells rendered permeable by virus infection and inhibiting translation, and as a selection agent for hygromycin resistance gene transformed cells. It is recommended for use as a selection agent at 100-800 μg/mL, specifically at 100 μg/mL for prokaryotes, 200 μg/mL for lower eukaryotes and 150-400 μg/mL for higher eukaryotes.

Biochem/physiol Actions

Mode of action: The product acts by inhibiting protein synthesis by inducing the misreading of the m-RNA template in the prokaryote, with the potency to inhibit translation.

Antimicrobial Spectrum: Hygromycin B acts against bacteria, fungi and higher eukaryotic cells.

Caution

Hygromycin B products should be stored as supplied at 2-8°C, and the dry solid is stable for at least 5 years if stored at 2-8°C. It is stable at 37°C for 30 days.

Preparation Note

This product is a solution that comes =60% (HPAE) and 45-60 mg/mL in H2O.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingqing Wang et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 35(8), 1582-1596 (2020-04-15)
Excessive osteoclast (OC) activity together with relatively weak osteoblast (OB) function are strongly connected to osteolytic diseases, including osteoporosis, tumor-induced osteolysis, and inflammatory bone erosion. Very few natural products or compounds have been shown to exert therapeutic effects on both
Rina Zuchman et al.
Journal of fungi (Basel, Switzerland), 7(2) (2021-02-04)
Protein phosphorylation cascades are universal in cell signaling. While kinome diversity allows specific phosphorylation events, relatively few phosphatases dephosphorylate key signaling proteins. Fungal mitogen activated protein kinases (MAPK), in contrast to their mammalian counterparts, often show detectable basal phosphorylation levels.
Yusheng Zhao et al.
Genes & development, 35(11-12), 888-898 (2021-05-15)
Plants monitor many aspects of their fluctuating environments to help align their development with seasons. Molecular understanding of how noisy temperature cues are registered has emerged from dissection of vernalization in Arabidopsis, which involves a multiphase cold-dependent silencing of the
Milena Szafraniec et al.
Dalton transactions (Cambridge, England : 2003), 41(32), 9728-9736 (2012-07-14)
Catalytic RNA molecules (ribozymes) have often been used for the testing of interactions of antibiotics with ribonucleic acids. We showed that the impact of capreomycin and hygromycin B on delta ribozyme catalysis might change dramatically, from stimulation to inhibition, depending
Xiang Liu et al.
Plant cell reports, 32(1), 77-87 (2012-11-01)
KEY MESSAGE : Efficient Agrobacterium -mediated genetic transformation for investigation of genetic and molecular mechanisms involved in inflorescence architectures in Cornus species. Cornus canadensis is a subshrub species in Cornus, Cornaceae. It has recently become a favored non-model plant species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service