Skip to Content
Merck
All Photos(1)

Key Documents

GW22549

Sigma-Aldrich

Anti-PPP1CC antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-22549

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1CC(5501)

Related Categories

Immunogen

Immunogen Sequence: GI # 4506007, sequence 1-323
Recombinant protein phosphatase 1, catalytic subunit, gamma isoform

Application

Anti-PPP1CC antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Serine/threonine-protein phosphatase PP1-γ catalytic subunit is an enzyme encoded by the PPP1CC gene in humans. A novel phosphatidic acid (PA) binding region on PP1c-γ is identified that contains a unique loop-strand structural fold responsible for the interaction with PA. PP1c γ may function as a mediator of PA action in cells. There may be an existence of a specific high affinity PA-binding domain on the enzyme. Its overexpression promotes vascular smooth muscle cell survival by interfering with JNK1 and p53 phosphorylation cascades involved in apoptosis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey A Jones et al.
Biochemistry, 44(40), 13235-13245 (2005-10-06)
Phosphatidic acid (PA) has been recognized as a lipid second messenger, yet few cellular targets for PA have been identified. Previous work demonstrated PA as a potent and noncompetitive tight-binding inhibitor of the catalytic subunit (gamma isoform) of protein phosphatase-1
Igor Tchivilev et al.
The Journal of biological chemistry, 283(32), 22193-22205 (2008-06-10)
The development of therapeutic strategies to inhibit reactive oxygen species (ROS)-mediated damage in blood vessels has been limited by a lack of specific targets for intervention. Targeting ROS-mediated events in the vessel wall is of interest, because ROS play important
Jeffrey A Jones et al.
The Journal of biological chemistry, 277(18), 15530-15538 (2002-02-22)
Phosphatidic acid (PA) has been identified as a bioactive lipid second messenger, yet despite extensive investigation, no cellular target has emerged as a mediator of its described biological effects. In this study, we identify the gamma isoform of the human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service