Skip to Content
Merck
All Photos(1)

Key Documents

GW21011

Sigma-Aldrich

Anti-β-2 Adrenergic Receptor antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-β-2 adrenergic receptor, Anti-β-2 adrenoreceptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21011

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRB2(154)

General description

β-2 Adrenergic Receptor is a member of the seven-transmembrane-domain receptor family. The ADRB2 gene is mapped on the human chromosome at 5q32. β-2 Adrenergic Receptor is a seven-transmembrane protein that consists of α helices. The N-terminal is extracellular whereas the C-terminal is intracellular.

Immunogen

Immunogen Sequence: GI # 4501969, sequence 335-408
Recombinant adrenergic, β-2-, receptor, surface

Application

Anti-β-2 Adrenergic Receptor antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The β-2 adrenergic receptor (β2 adrenoreceptor), also known as ADRB2, is present within a cell membrane. It reacts with adrenaline (epinephrine) as a hormone or neurotransmitter affecting muscles or organs. It may be a risk factor for chronic pain and its polymorphisms may influence labor pain. It contains a common non-synonymous single nucleotide polymorphism. Epinephrine activates ADRB2 impairing keratinocyte migration. β2-adrenoceptor pathway is essential for cardiovascular regulation.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis W McGraw et al.
Proceedings of the American Thoracic Society, 2(4), 292-296 (2005-11-04)
It is now clear that the beta2-adrenergic receptor continuously oscillates between various conformations in the basal state, and that agonists act to stabilize one or more conformations. It is conceivable that synthetic agonists might be engineered to preferentially confine the
R A Hall et al.
Nature, 392(6676), 626-630 (1998-04-29)
Stimulation of beta2-adrenergic receptors on the cell surface by adrenaline or noradrenaline leads to alterations in the metabolism, excitability, differentiation and growth of many cell types. These effects have traditionally been thought to be mediated exclusively by receptor activation of
U H Frey et al.
British journal of anaesthesia, 112(3), 477-484 (2013-12-25)
While the β2-adrenoceptor pathway is essential for cardiovascular regulation, the impact of ADRB2 gene variations on circulatory responses is unclear, possibly due to neural compensatory mechanisms. We tested the hypotheses that (i) sympathetic block by thoracic epidural anaesthesia (TEA) unmasks
Yongjun He et al.
Oncotarget, 8(11), 18206-18212 (2017-02-18)
High altitude pulmonary edema (HAPE) occurs mainly under conditions such as high altitude, rapid ascent, or hypoxia. Previous studies suggest that ADRB2, GNB3, TH, and GSTP1 polymorphisms are associated with various lung diseases. We evaluated whether those polymorphisms are associated
Victor E Ortega et al.
The Lancet. Respiratory medicine, 2(3), 204-213 (2014-03-14)
Severe adverse life-threatening events associated with longacting β agonist (LABA) use have caused the US Food and Drug Administration (FDA) to review the safety of these drugs, resulting in a boxed warning and a mandatory safety study in 46 800 patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service