Skip to Content
Merck
All Photos(2)

Key Documents

GHS216

Sigma-Aldrich

Hematoxylin Solution, Gill No. 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

4 g/L

pH

2.5-3.5

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Gill formulation No. 2 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.

Other Notes

4 g/L certified hematoxylin

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haley R Noonan et al.
Disease models & mechanisms, 9(8), 873-884 (2016-08-05)
Patients with von Hippel-Lindau (VHL) disease harbor a germline mutation in the VHL gene leading to the development of several tumor types including clear cell renal cell carcinoma (ccRCC). In addition, the VHL gene is inactivated in over 90% of
Yitzhak Reizel et al.
Genes & development, 34(15-16), 1039-1050 (2020-06-21)
The FoxA transcription factors are critical for liver development through their pioneering activity, which initiates a highly complex regulatory network thought to become progressively resistant to the loss of any individual hepatic transcription factor via mutual redundancy. To investigate the
G Gonzalez et al.
Diseases of the esophagus : official journal of the International Society for Diseases of the Esophagus, 29(6), 670-680 (2015-08-08)
Deep esophageal glands play a vital role in the protection and regeneration of the esophageal mucosa. Conditions such as gastroesophageal reflux disease and Barrett's esophagus have been associated with a change in the usual glands by oncocytic metaplasia. However, little
Irene Anastasia et al.
Cell reports, 34(11), 108873-108873 (2021-03-18)
Contacts between organelles create microdomains that play major roles in regulating key intracellular activities and signaling pathways, but whether they also regulate systemic functions remains unknown. Here, we report the ultrastructural organization and dynamics of the inter-organellar contact established by
Noora Puhakka et al.
PloS one, 12(3), e0172521-e0172521 (2017-03-09)
Traumatic brain injury (TBI) can result in several dentate gyrus-regulated disabilities. Almost nothing is known about the chronic molecular changes after TBI, and their potential as treatment targets. We hypothesized that chronic transcriptional alterations after TBI are under microRNA (miRNA)

Related Content

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service