Documents
GE28-4064-10
Sigma-Aldrich
Tricorn 5/100 Column
Synonym(s):
5/100 Tricorn Column, Tricorn Column
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
material
glass column
packaging
1 ea of
General description
Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Tricorn columns include a glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock, and filter holder.
Features and Benefits
Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Visit the Document Library
Articles
Capto MMC is a multimodal cation exchanger with the properties of a weak cation exchanger. In addition to electrostatic interactions, the ligand structure provides for additional interaction modes such as hydrophobic interaction, hydrogen bonding, and thiophilic interaction.
This page describes the difference between bind/elute and flowthrough modes in multimodal chromatography using Cytiva products.
Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.
Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.
See All
Protocols
This page covers using SOURCE™ medias for the purification of proteins, peptides, or oligonucleotides.
This page covers how and when to use Sepharose XL media for purification of proteins.
Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.
This page discusses various aspects of sample preparation for chromatographic purification.