Skip to Content
Merck
All Photos(1)

Key Documents

G9534

Sigma-Aldrich

Globotriaosylsphingosine from porcine blood

Synonym(s):

α-D-Gal-(1→4)-β-D-Gal-(1→4)-β-D-Glc-1→O-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H67NO17
CAS Number:
Molecular Weight:
785.91
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

lipid type

sphingolipids

Quality Level

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\C(O)C(N)COC1OC(CO)C(OC2OC(CO)C(OC3OC(CO)C(O)C(O)C3O)C(O)C2O)C(O)C1O

InChI

1S/C36H67NO17/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-21(41)20(37)19-49-34-30(47)27(44)32(23(17-39)51-34)54-36-31(48)28(45)33(24(18-40)52-36)53-35-29(46)26(43)25(42)22(16-38)50-35/h14-15,20-36,38-48H,2-13,16-19,37H2,1H3/b15-14+

InChI key

GRGNVOCPFLXGDQ-CCEZHUSRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henrik Gold et al.
Clinical chemistry, 59(3), 547-556 (2012-12-15)
Biochemical markers that accurately reflect the severity and progression of disease in patients with Fabry disease and their response to treatment are urgently needed. Globotriaosylsphingosine, also called lysoglobotriaosylceramide (lysoGb3), is a promising candidate biomarker. We synthesized lysoGb3 and isotope-labeled [5,6,7,8,9]
Shigeki Sugawara et al.
Biochimica et biophysica acta, 1790(2), 101-109 (2008-11-18)
Heat shock proteins (HSPs) are divided into stress-inducible and constitutive types. Generally, HSP70 (stress inducible) and HSC70 (constitutive) are representative of their types, respectively. From the results of immunocytochemical analysis, both HSP70 and HSC70 were constitutively expressed in globotriaosylceramide (Gb3)-expressing
Christiane Auray-Blais et al.
Analytical chemistry, 84(6), 2745-2753 (2012-02-09)
Fabry disease is a lysosomal storage disorder caused by deficiency of α-galactosidase A, resulting in glycosphingolipid accumulation in organs and tissues, including plasma and urine. Two disease-specific Fabry biomarkers have been identified and quantified in plasma and urine: globotriaosylceramide (Gb(3))
Michel Boutin et al.
Clinica chimica acta; international journal of clinical chemistry, 414, 273-280 (2012-10-09)
Fabry disease is a complex, multisystemic and clinically heterogeneous disease, with elevated excretion of globotriaosylceramide (Gb(3)) and globotriaosylsphingosine (lyso-Gb(3)) accumulating in biological fluids caused by deficiency of the enzyme, lysosomal α-galactosidase A. Our aims were to propose a tandem mass
Mariëlle J van Breemen et al.
Biochimica et biophysica acta, 1812(1), 70-76 (2010-09-21)
Fabry disease is treated by two-weekly infusions with α-galactosidase A, which is deficient in this X-linked globotriaosylceramide (Gb3) storage disorder. Elevated plasma globotriaosylsphingosine (lysoGb3) is a hallmark of classical Fabry disease. We investigated effects of enzyme replacement therapy (ERT) on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service