Skip to Content
Merck
All Photos(2)

Key Documents

G8255

Sigma-Aldrich

Glutamic-Pyruvic Transaminase from porcine heart

lyophilized powder, ≥75 units/mg protein

Synonym(s):

L-Alanine: 2-Oxoglutarate Aminotransferase, Alanine Aminotransferase (catalytic concentration), GPT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
Beilstein:
3562140
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Porcine heart

Quality Level

form

lyophilized powder

specific activity

≥75 units/mg protein

composition

Protein, 75-95%

technique(s)

cell culture | mammalian: suitable

color

light yellow to yellow

foreign activity

L-glutamic dehydrogenase, lactic dehydrogenase and malic dehydrogenase ≤0.01%
glutamic-oxalacetic transaminase ≤0.1%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Glutamic-pyruvic transaminase (GPT) is localized majorly in the liver.

Research area: Cell Signaling

Application

Glutamic-Pyruvic Transaminase from porcine heart has been used:
  • in the transamination assay with a racemic mixture of synthetic 3-methyl-2-oxoglutarate
  • to remove glutamate from the cell culture medium used for culturing human embryonic kidney (HEK293) cells
  • in the lactate-dehydrogenase (LD)-coupled alanine transaminase (ALT) reaction kinetic studies

Biochem/physiol Actions

Glutamic-pyruvic transaminase (GPT) mediates the transfer of the amino group to α-ketoglutarate from l-alanine which results in pyruvate and glutamate.Thereby GPT plays a key role in gluconeogenesis and amino acid metabolism. The role of GPT in various kinds of tumors has been reported.
Glutamic-pyruvic transaminase (GPT) is located in most cells in the body, but is found in highest concentrations in the liver. It is used as a marker of liver disease due to the release of GPT into the blood following liver cell necrosis.

Unit Definition

One unit will convert 1.0 μmole of α-ketoglutarate to L-glutamate per min at pH 7.6 at 37 °C in the presence of L-alanine.

Physical form

Lyophilized powder containing citrate and acetate

Storage and Stability

Tightly closed. Dry. Keep locked up or in an area accessible only to qualified or authorized
persons

Analysis Note

Protein determined by biuret

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Mahlert et al.
Journal of the American Chemical Society, 129(39), 12011-12018 (2007-09-06)
The acidic lipopeptides, including the calcium-dependent antibiotics (CDA), daptomycin, and A54145, are important macrocyclic peptide natural products produced by Streptomyces species. All three compounds contain a 3-methyl glutamate (3-MeGlu) as the penultimate C-terminal residue, which is important for bioactivity. Here
Wei Guo et al.
Cancers, 12(7) (2020-07-15)
Metabolic reprogramming is an essential hallmark of cancer. Besides the "Warburg effect", cancer cells also actively reprogram amino acid metabolism to satisfy high nutritional demands in a nutrient-poor environment. In the glucose-alanine cycle, exogenous alanine taken up by hepatocytes is
A simple and sensitive detection of glutamic-pyruvic transaminase activity based on fluorescence quenching of bovine serum albumin
Chen M, et al.
Royal Society of Chemistry Advances, 5, 103557-103562 (2015)
A Comprehensive Guide to Toxicology in Nonclinical Drug Development
Faqi A S
Biomarkers journal., 2017 null
Giordano Pula et al.
Journal of biochemical and biophysical methods, 64(3), 167-181 (2005-08-23)
The surface expression of G protein-coupled receptors is regulated by internalization. For many receptors, a constitutive level of internalization in the absence of agonist has been reported. The constitutive internalization of metabotropic glutamate receptor 1a (mGluR1a) has been described, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service