Skip to Content
Merck
All Photos(1)

Key Documents

G8160

Sigma-Aldrich

Granulocyte Colony-Stimulating Factor from mouse

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

G-CSF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Granulocyte Colony-Stimulating Factor from mouse, G-CSF, recombinant, expressed in E. coli, suitable for cell culture

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

10-60 pg/mL ED50

quality

endotoxin tested

mol wt

19 kDa (179 amino acids including N-terminal methionine)

packaging

pkg of 5 and 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤0.01 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

The Csf3r (colony stimulating factor 3 receptor (granulocyte)) gene encodes a prototypical mobilizing cytokine.[1]

Application

Granulocyte Colony-Stimulating Factor from mouse has been used in cytokine therapy.[2]

Biochem/physiol Actions

G-CSF is a proliferation, differentiation, survival, and activation factor for hematopoietic cells of the restricted neutrophilic granulocyte lineage. It is produced by macrophages activated by endotoxin (LPS), by monocytes activated by TNF-α with IFNγ, by fibroblasts and endothelial cells activated by IL-1 or TNF-α, and by bone marrow stromal cells activated by IL-1 or LPS. It is suggested that during the inflammatory process endotoxins stimulate tissue macrophages to produce not only G-CSF but several other cytokines, including IL-1 and TNF-α, which in turn stimulate more G-CSF release from endothelial cells and fibroblasts. G-CSF can also synergize with IL-3 to shorten the G0 period of early hematopoietic progenitors. In addition to the namesake proliferative activity, G-CSF acts on mature neutrophils to enhance their survival and to stimulate their tumorcidal activity. Human G-CSF binds and activates a 130 kD to150 kD glycoprotein single chain receptor that has been classified as a member of the hematopoietic (cytokine) receptor family, the cytokine receptor class I, or the gp130 related cytokine receptor family (although it does not apparently bind to gp130). G-CSF receptors can be found on neutrophils, myeloid leukemia cells that respond to G-CSF, bone marrow cells of neutrophilic granulocyte lineage, and on placental trophoblasts, plus a soluble form may be expressed. Two forms of human G-CSF (177 and 174 amino acids) are synthesized from a single gene by alternative splicing, but murine G-CSF is a single expressed form of 178 amino acids. Human and murine G-CSF share 73% amino acid sequence homology and full cross-reactivity.
Granulocyte colony-stimulating factor (G-CSF) is a lineage-restricted hematopoietic growth factor, stimulating final mitotic divisions and the terminal cellular maturation of the partially differentiated hematopoietic progenitors. G-CSF3,4 is produced by monocytes and fibroblasts. It stimulates granulocyte colony formation, activates neutrophils and mature granulocytes, and promotes differentiation of certain myeloid leukemic cells. Natural G-CSF is a glycoprotein of 177 amino acids and a molecular weight of 19 kDa.5 Human and murine G-CSF have about 75% homology and show biological cross-reactivity.

Physical form

Lyophilized from a 0.2 μm filtered solution in 10 mM citric acid, 0.01% TWEEN® 20, pH 4.0, containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured in a cell proliferation assay using a murine myeloblastic cell line, NFS-60.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combining pharmacological mobilization with intramyocardial delivery of bone marrow cells over-expressing VEGF is more effective for cardiac repair
Wang Y, et al.
Journal of Molecular and Cellular Cardiology, 40(5), 736-745 (2006)
Expression of the G-CSF receptor in monocytic cells is sufficient to mediate hematopoietic progenitor mobilization by G-CSF in mice
Christopher MJ, et al.
The Journal of Experimental Medicine (2011)
Hitesh S Deshmukh et al.
Nature medicine, 20(5), 524-530 (2014-04-22)
Neonatal colonization by microbes, which begins immediately after birth, is influenced by gestational age and the mother's microbiota and is modified by exposure to antibiotics. In neonates, prolonged duration of antibiotic therapy is associated with increased risk of late-onset sepsis
Expression of the G-CSF receptor in monocytic cells is sufficient to mediate hematopoietic progenitor mobilization by G-CSF in mice
ChristopherMJ, et al.
The Journal of Experimental Medicine (2011)
Rui Zhang et al.
Hematology (Amsterdam, Netherlands), 20(10), 555-560 (2015-06-13)
The treatment for acute myeloid leukemia (AML) remains an important clinical problem. Recently, hematopoietic stem cell therapy provides promising outcomes. In this study we examined the clinical effect of HLA-mismatched hematopoietic stem cell infusion combined with chemotherapy as a postremission

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service