Skip to Content
Merck
All Photos(1)

Key Documents

G8021

Sigma-Aldrich

Monoclonal Anti-β-Galactosidase antibody produced in mouse

clone GAL-13, ascites fluid

Synonym(s):

Monoclonal Anti-β-Galactosidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

GAL-13, monoclonal

contains

15 mM sodium azide

species reactivity

bacteria

technique(s)

dot blot: suitable using soluble β-galactosidase
immunocytochemistry: suitable
indirect ELISA: 1:2,000 using mouse primary antibody, bridging antibody and Escherichia coli β-galactosidase

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Escherichia coli ... lacZ(1036228) , lacZ(1036228)
Escherichia coli K12 ... Lacz(945006)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-β-Galactosidase (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. β −galactosidase is a E coli protein and is a tetramer with 1023 amino acids.

Specificity

The antibody may be used for amplification in immunoenzymatic staining by preparing a β-galactosidase anti-β-galactosidase (BGABG) soluble complex. It is not recommended for immunoblotting; it does not recognize denatured or reduced β-galactosidase.

Immunogen

β-galactosidase from E. coli

Application

Monoclonal Anti-β-Galactosidase antibody produced in mouse has been used in:

  • immunostaining and histochemistry
  • immunostaining and microscopy
  • immunohistochemistry
  • immunofluorescence staining
  • enzyme linked immunosorbent assay(ELISA)
  • immuno dot blot

Biochem/physiol Actions

β −galactosidase substrate is lactose. Synthetic substrates like o-nitrophenyl-β-d-galactopyranoside (oNPG) or p-nitrophenyl-β-d-galactopyranoside (pNPG) are used in enzymatic assays. It is also used as marker in studying gene expression. The use of monoclonal β -galactosidase anti- β -galactosidase (BGABG) soluble complex in the unlabeled antibody-enzyme method results in an intense signal with a very low background.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thyroid hormone signaling in the mouse retina
Arbogast P, et al.
PLoS ONE, 11(12), e0168003-e0168003 (2016)
Shuang Li et al.
Science signaling, 7(332), ra62-ra62 (2014-07-06)
Hedgehog (Hh) is a secreted glycoprotein that binds its receptor Patched to activate the G protein (heterotrimeric guanine nucleotide-binding protein)-coupled receptor-like protein Smoothened (Smo). In Drosophila, protein kinase A (PKA) phosphorylates and activates Smo in cells stimulated with Hh. In
Heng Zhao et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 24(6), 681-692 (2004-06-08)
Apoptosis plays a critical role in many neurologic diseases, including stroke. Cytochrome c release and activation of various caspases are known to occur after focal and global ischemia. However, recent reports indicate that caspase-independent pathways may also be involved in
Transcription factor TLX1 controls retinoic acid signaling to ensure spleen development
Lenti E, et al.
The Journal of Clinical Investigation, 126(7), 2452-2464 (2016)
Anja Weimer et al.
Molecular medicine (Cambridge, Mass.), 18, 346-358 (2011-12-14)
Administration of therapeutic genes to human osteoarthritic (OA) cartilage is a potential approach to generate effective, durable treatments against this slow, progressive disorder. Here, we tested the ability of recombinant adeno-associated virus (rAAV)-mediated overexpression of human insulinlike growth factor (hIGF)-I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service