Skip to Content
Merck
All Photos(1)

Key Documents

G4420

Sigma-Aldrich

Anti-GRP94 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Endoplasmin precursor, Anti-Glucose Regulated Protein 94, Anti-Tumor Rejection Antigen 1, Anti-gp96

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 94 kDa

species reactivity

canine, human, mouse

concentration

~1 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): 10-20 μg using MDCK whole cell lysate
indirect immunofluorescence: 2-4 μg/mL using HeLa cells
microarray: suitable
western blot: 0.5-1 μg/mL using whole cell extracts of HeLa, MDCK, and NIH3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSP90B1(7184)
mouse ... Hsp90b1(22027)
rat ... Tra1(298957)

Immunogen

synthetic peptide corresponding to amino acids 733-750 located near the C-terminus of human GRP94. This sequence is identical in mouse, rat, dog, porcine, and bovine GRP94 and highly conserved (1 amino acid substitutuion) in chicken.

Application

Anti-GRP94 (C-terminal) antibody produced in rabbit has been used in immunocytochemistry.
Anti-GRP94 (C-terminal) antibody produced in rabbit is suitable for immunoblotting at a working concentration of 0.5-1μg/mL using whole cell extracts of the human epitheloid carcinoma HeLa cells, Madin Darby canine kidney (MDCK) cells, and mouse fibroblast NIH3T3 cells. It is also suitable for immunoprecipitation of GRP94 protein from a MDCK whole cell lysate at 10-20μg concentration.

Biochem/physiol Actions

GRP94 (glucose-regulated protein 94) is involved in protein folding, sorting and secretion, and binding of immunogenic peptides. GRP94 has a role in peptide antigen presentation to major histocompatibility complex class I molecules of antigen presenting cells (APCs). It binds specifically to CD91 (α2-macroglobulin receptor), and possibly to the Toll-like receptors (TLRs) on the surface of APCs. Overexpression of GRP94 has been associated with cellular transformation and tumorigenesis.
GRPs are unresponsive to heat stress and are induced by stress related to glucose starvation or defects in glycoprotein processing, depletion of Ca2+ stores, acidosis, and hypoxia conditions that are also common in poorly vascularized tumor tissues.
The GRP94 (glucose-regulated protein 94) is a 94 kDa Ca2+- binding glycoprotein that belongs to a subfamily of the heat shock proteins Hsp90. It is a chaperone protein constitutively localized to the endoplasmic reticulum (ER) of mammalian cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ménoret et al.
International journal of cancer, 56(3), 400-405 (1994-02-01)
Resistance to glucose starvation and expression of glucose-regulated proteins (grp) were studied in a model of rat colon carcinoma. In this model, various clones originating from the same parental tumor showed distinct tumorigenic potential in syngeneic hosts. Some clones were
Y Argon et al.
Seminars in cell & developmental biology, 10(5), 495-505 (1999-12-22)
GRP94 is the ER representative of the HSP90 family of stress-induced proteins. It binds to a limited number of proteins in the secretory pathway, apparently by recognizing advanced folding intermediates or incompletely assembled proteins, GRP94 also binds peptides and can
P K Srivastava et al.
Current opinion in immunology, 6(5), 728-732 (1994-10-01)
Heat shock proteins (HSPs) are associated with a broad spectrum of peptides derived from the cells from which they are isolated. Vaccination with such HSP-peptide complexes elicits protective immunity against tumors or other cells used as the source of HSPs.
R J Binder et al.
Nature immunology, 1(2), 151-155 (2001-03-15)
Antigen presenting cells (APCs) can take up exogenous antigenic peptides chaperoned by heat shock protein gp96 and re-present them through the endogenous pathway on their major histocompatibility class I molecules. The high efficiency of this process has been attributed previously
Ramachandra K Reddy et al.
Cancer research, 62(24), 7207-7212 (2002-12-25)
A major challenge in treating cancer is the difficulty of bringing therapy to poorly perfused areas of solid tumors, which are often most resistant to chemotherapy and radiation. GRP94 is a chaperone protein localized in the endoplasmic reticulum with antiapoptotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service