Skip to Content
Merck
All Photos(1)

Key Documents

G4252

Sigma-Aldrich

Glyoxalase I from Saccharomyces cerevisiae

Grade IV, buffered aqueous glycerol solution, ≥400 units/mg protein

Synonym(s):

S-Lactoyl-glutathione methylglyoxal-lyase (isomerizing)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Grade IV

Quality Level

form

buffered aqueous glycerol solution

specific activity

≥400 units/mg protein

foreign activity

glyoxalase II ≤1%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Glyoxalase detoxification system consists of glyoxalase (GLO)-I and GLO-II. GLO-I is a cytosolic, 42 kDa, dimeric Zn2+ metalloenzyme.

Application

Glyoxalase I from Saccharomyces cerevisiae has been used as a standard in Glyoxalase I assay for C. dilatata females and brooded encapsulated embryos. It has also been used as a standard for calibration curve generation for quantifying glyoxalase I from brain tissues and cerebral microvessels.

Biochem/physiol Actions

Glyoxalase I is universally expressed and involved in the protection against cellular damage due to cytotoxic metabolites such as advanced glycation end products (AGEs). It is an integral component of the detoxification system, catalyzing the conversion of reactive, acyclic a-oxoaldehydes into the corresponding a-hydroxyacids in a glutathione-dependent manner.

Unit Definition

One unit will form 1.0 μmole of S-lactoylglutathione from methylglyoxal and reduced glutathione per min at pH 6.6 at 25 °C.

Physical form

Solution in 50% glycerol, 0.4 M (NH4)2SO4 and 0.002 M KH2PO4 pH 6.5

Analysis Note

Protein determined by biuret.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unicameral bone cyst in the calcaneus with pathologic fracture. A literature review and case report.
N A Grumbine et al.
Journal of the American Podiatric Medical Association, 76(2), 96-99 (1986-02-01)
Víctor Cubillos et al.
Marine environmental research, 119, 59-71 (2016-05-28)
The estuarine slipper limpet Crepipatella dilatata is a gastropod that can survive prolonged periods of low salinities (< 24 PSU) caused by tidal changes and/or prolonged periods of rain. During low salinity events, C. dilatata can isolate its body from the outside
Hiroya Taniguchi et al.
Molecular cancer therapeutics, 11(10), 2294-2300 (2012-07-13)
Methylglyoxal is an essential component in glycolysis and is known to be an inducer of apoptosis. Glyoxalase I (GLO1) metabolizes and inactivates methylglyoxal. GLO1 is known to be overexpressed in cancer cells and causes resistance to anticancer agents. We show
Lingling Dong et al.
The journal of obstetrics and gynaecology research, 38(8), 1077-1085 (2012-05-01)
Metformin plays an important role in the inhibition of cancer cell growth and prolongs remission durations. It reverses progestin-resistance in endometrial cancer cells by downregulating glyoxalase I (GloI) expression. This study aimed to investigate the effect of metformin on endometrial
Ravi Gupta et al.
Journal of proteome research, 11(5), 2684-2696 (2012-04-11)
Plants' distribution and productivity are adversely affected by low temperature (LT) stress. LT induced proteins were analyzed by 2-DE-nano-LC-MS/MS in shoot secretome of Hippophae rhamnoides (seabuckthorn), a Himalayan wonder shrub. Seedlings were subjected to direct freezing stress (-5 °C), cold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service