Skip to Content
Merck
All Photos(1)

Key Documents

G2299

SAFC

L-Glutathione oxidized

Pharma Manufacturing

Synonym(s):

GSSG, Glutathiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N6O12S2
CAS Number:
Molecular Weight:
612.63
Beilstein:
1718700
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

solubility

H2O: 10 mg/mL, clear, colorless

suitability

suitable for manufacturing use

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10-,11-,12-/m0/s1

InChI key

YPZRWBKMTBYPTK-BJDJZHNGSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glu-Cys-Gly, Glu-Cys-Gly [Disulfide bridge: 2a-2b]

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun-Peng Wang et al.
Molecular medicine reports, 9(5), 1813-1819 (2014-03-08)
Schisandra chinensis is a well-known traditional medicinal herb. Acetaminophen is a commonly used over-the-counter analgesic and overdose of acetaminophen was the most frequent cause of acute liver failure. However, no studies have demonstrated the role of Schisandra chinensis in acetaminophen-induced
Fabio Di Domenico et al.
Expert review of neurotherapeutics, 15(1), 19-40 (2014-09-23)
Alzheimer's disease (AD), characterized by progressive loss of memory, language, reasoning and other cognitive functions, including dementia, is characterized pathologically by the presence of senile plaques, neurofibrillary tangles and synapse loss. Increased oxidative/nitrosative stress, decreased antioxidants, mitochondrial damage and other
Vladimir L Kolossov et al.
Experimental biology and medicine (Maywood, N.J.), 239(4), 394-403 (2014-03-04)
The glutathione couple GSH/GSSG is the most abundant cellular redox buffer and is not at equilibrium among intracellular compartments. Perturbation of glutathione poise has been associated with tumorigenesis; however, due to analytical limitations, the underlying mechanisms behind this relationship are
N Mohell et al.
Cell death & disease, 6, e1794-e1794 (2015-06-19)
Two main causes of platinum resistance are mutation in the tumor suppressor gene TP53 and drug-induced increase in intracellular glutathione concentration. Mutations in TP53 occur in about 50% of human tumors. APR-246 (PRIMA-1(MET)) is the first clinical-stage compound that reactivates
Changchun Cai et al.
Hepatology (Baltimore, Md.), 60(3), 990-1002 (2014-05-07)
Acetaminophen (APAP) toxicity is the most common cause of acute liver failure in industrialized countries. Understanding the mechanisms of APAP-induced liver injury as well as other forms of sterile liver injury is critical to improve the care of patients. Recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service